Skip to Content
Merck
All Photos(1)

Key Documents

04476

Sigma-Aldrich

Hordenine

≥97.0% (HPLC)

Synonym(s):

2-(4-Hydroxyphenyl)-N,N-dimethylethylamine, 4-(2-Dimethylaminoethyl)phenol, N,N-Dimethyltyramine, p-Hydroxy-N,N-dimethylphenethylamine, Anhaline, Cactine, Eremursin, Peyocactine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Beilstein:
2207615
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97.0% (HPLC)

form

powder or crystals

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

CN(C)CCc1ccc(O)cc1
CN(C)CCc1ccc(O)cc1

InChI

1S/C10H15NO/c1-11(2)8-7-9-3-5-10(12)6-4-9/h3-6,12H,7-8H2,1-2H3

InChI key

KUBCEEMXQZUPDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hordenine is suitable to study the effect of reed canary grass alkoloids on in vito digestibility. It is also suitable for use as a standard to identify endogenous hordenine in ungerminated and germinated barley by HPLC/diode array detection analysis. Hordenine may be used as an analytical reference material and in research on the activities of phenethylamine type alkaloids.

Biochem/physiol Actions

Hordenine is an alkaloid found in plants, e.g. the roots of germinating barley and marine algae. It is a metabolite in tyrosine metabolism, biosynthesized from tyramine by two subsequent N-methylations, it is metabolized by monoamine oxidase. Hordenine is a sympathomimetic and its pharmacological actions are of interest, as it is occasionally found in post race urine samples. Hordenine inhibits melanogenesis by suppression of cyclic adenosine monophosphate (cAMP) production. cAMP is involved in the expression of melanogenesis-related proteins. Hordenine may be used in the inhibition of hyperpigmentation. It is present in barley roots from the first day of seed germination, but is not present in seeds.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hordenine content of the marine alga Mastocarpus stellatus and the algal food product carrageen.
Barwell, C.J., et al.
Phytotherapy Research, 3, 67-69 (1989)
Studies on the mechanism of action of monoamine oxidase: metabolism of N,N-dimethyltryptamine and N,N-dimethyltryptamine-N-oxide.
T E SMITH et al.
Biochemistry, 1, 137-143 (1962-01-01)
A K Singh et al.
Forensic science international, 54(1), 9-22 (1992-04-01)
Hordenine cross-reacted with various enzyme linked immunosorbent assay (ELISA) or radioimmunoassay (RIA) kits used for the screening of urine samples. Morphine-ELISA kit was most sensitive, whereas etorphine- and buprenorphine-ELISA kits were least sensitive to hordenine cross-reactivity. Hordenine also interfered with
C A Bourke et al.
Australian veterinary journal, 65(7), 218-220 (1988-07-01)
The acute toxicity for sheep of 3 alkaloids that occur in Phalaris acquatica was examined by intravenous and oral administration. The lowest tested dose rates that produced clinically observed signs were, for 5-methoxy dimethyltryptamine, 0.1 mg/kg body weight intravenously and
C J Barwell et al.
The Journal of pharmacy and pharmacology, 41(6), 421-423 (1989-06-01)
The selectivity of the naturally occurring amine, N,N-dimethyltyramine (hordenine) for monoamine oxidase (MAO) and its action upon isolated vasa deferentia of the rat was investigated. Hordenine was deaminated by rat liver MAO with a Michaelis constant of 479 microM and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service