Skip to Content
Merck
All Photos(1)

Key Documents

T90344

Sigma-Aldrich

Tyramine

≥98.0%

Synonym(s):

2-(4-Hydroxyphenyl)ethylamine, 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
1099914
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

bp

175-181 °C/8 mmHg (lit.)

mp

160-162 °C (lit.)

SMILES string

NCCc1ccc(O)cc1

InChI

1S/C8H11NO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6,9H2

InChI key

DZGWFCGJZKJUFP-UHFFFAOYSA-N

Gene Information

rat ... Drd2(24318)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Tyramine is extensively used in the preparation of a variety of hydrogels for biomedical applications.
  • It is used as a key precursor in the total synthesis of (−)-mesembrine and (−)-galanthamine.
  • It can also be used in the preparation of tyramine-functionalized graphene quantum dots (GQDs) as fluorescence reporters for optical sensing of metabolites.

Biochem/physiol Actions

Can enter catecholaminergic terminals and be released as a false transmitter.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Total Synthesis of (−)-Galanthamine by Remote Asymmetric Induction.
Kodama, Sumiaki et al.
Angewandte Chemie (International Edition in English), 43(20), 2659-2661 (2004)
Ultrasensitive profiling of metabolites using tyramine-functionalized graphene quantum dots.
Li N, et al.
ACS Nano, 10(3), 3622-3629 (2016)
Desymmetrization of cyclohexadienones via cinchonine derived thiourea-catalyzed enantioselective aza-Michael reaction and total synthesis of (−)-Mesembrine
Gu Q and You S-L
Chemical Science, 2(8), 1519-1522 (2011)
Enzyme-mediated fast in situ formation of hydrogels from dextran-tyramine conjugates.
Jin R, et al.
Biomaterials, 28(18), 2791-2800 (2007)
Simona Distinto et al.
European journal of medicinal chemistry, 48, 284-295 (2012-01-10)
Monoamine oxidase B (MAO-B) is a promising target for the treatment of neurodegenerative disorders. We report the synthesis and the biological evaluation of halogenated derivatives of 1-aryliden-2-(4-phenylthiazol-2-yl)hydrazines. The fluorinated series shows interesting activity and great selectivity toward the human recombinant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service