Skip to Content
Merck
All Photos(1)

Key Documents

00017

Sigma-Aldrich

Acacetin

≥97.0% (HPLC)

Synonym(s):

4′-Methylapigenin, 5,7-Dihydroxy-4′-methoxyflavone, Apigenin 4′-methyl ether, Buddleoflavonol, Linarigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Beilstein:
277879
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32150233
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (HPLC)

form

powder or crystals

mp

260-265 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2
COc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H12O5/c1-20-11-4-2-9(3-5-11)14-8-13(19)16-12(18)6-10(17)7-15(16)21-14/h2-8,17-18H,1H3

InChI key

DANYIYRPLHHOCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acacetin belongs to the category of naturally occurring plant pigments called flavonoids found in vascular plants. It was first reported to be extracted from the leaves of Robinia pseuducacia. It has been used as an active component of traditional Chinese medicine Xuelianhua.[1][2]

Biochem/physiol Actions

Acacetin has been noted to exhibit anti-peroxidative, anti-inflammatory, and anti-plasmodial properties. It has been posited to prevent the proliferation of Hep G2 cells, thereby causing cell apoptosis and subsequent anti-cancer action.[3] It has antiarrhythmic properties and can be used in the treatment of Atrial fibrillation.[2]
Flavonoid with anti-peroxidative and anti-inflammatory properties. Used in traditional Chinese medicine.
Flavonoid with anti-peroxidative and anti-inflammatory properties. Used in traditional Chinese medicine. It has been recognized as an antiproliferative agent on cancer cell lines by inducing apoptosis and blocking cell cycle progression in the G1 phase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acacetin, a natural flavone, selectively inhibits human atrial repolarization potassium currents and prevents atrial fibrillation in dogs
Li GR, et al.,
Circulation, 117(19), 2449-2457 (2008)
V Zimkhitha Sotenjwa et al.
Fitoterapia, 147, 104740-104740 (2020-10-12)
Artemisia afra (African wormwood) is a popular medicinal plant of southern Africa and is an excellent candidate for commercialisation. This current study was aimed at exploring the phytochemistry and chemical variation of non-volatile compounds within wild populations of A. afra
Acacetin inhibits the proliferation of Hep G2 by blocking cell cycle progression and inducing apoptosis
Hsu YL, et al.,
Biochemical Pharmacology, 67(5), 823-829 (2004)
Antifungal and antioxidant activities of heartwood, bark, and leaf extracts of Robinia pseudoacacia
Hosseinihashemi SK, et al.
BioResources, 11(1), 1634-1646 (2016)
Hui-Jun Wu et al.
Journal of molecular and cellular cardiology, 51(6), 966-973 (2011-09-13)
We have demonstrated that the natural flavone acacetin selectively inhibits ultra-rapid delayed rectifier potassium current (I(Kur)) in human atria. However, molecular determinants of this ion channel blocker are unknown. The present study was designed to investigate the molecular determinants underlying

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service