Skip to Content
Merck
All Photos(4)

Documents

M0631

Sigma-Aldrich

Magnesium acetate tetrahydrate

≥99%, ReagentPlus®

Synonym(s):

Acetic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45
Beilstein:
3730605
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
39021909
PubChem Substance ID:
NACRES:
NA.21

product line

ReagentPlus®

Assay

≥99%

form

powder
solid

mp

72-75 °C (lit.)

SMILES string

O.O.O.O.CC(=O)O[Mg]OC(C)=O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium acetate tetrahydrate can be used as:
  • A catalyst in the methanolysis of polylactic acid (PLA) to produce methyl lactate.
  • A reagent in the preparation of homochiral metal-organic frameworks by using tartrate ligands.
  • A precursor to synthesize magnesium oxide (MgO) fillers for polymer composites.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and investigation of SiO2-MgO coated MWCNTs and their potential application
Nemeth K, et al.
Scientific Reports, 9, 1-11 (2019)
Homochiral frameworks derived from magnesium, zinc and copper salts of L-tartaric acid
Rood J, et al.
Journal of Solid State Chemistry, 183, 270-276 (2010)
Methanolysis of Poly (lactic Acid) Using Catalyst Mixtures and the Kinetics of Methyl Lactate Production
Lamberti F, et al.
Polymers, 14, 1763-1763 (2022)
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service