Skip to Content
Merck
All Photos(5)

Key Documents

C4901

Sigma-Aldrich

Calcium chloride

anhydrous, powder, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥97%
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Agency

suitable for SM 5210

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥97%

form

powder

mp

772 °C (lit.)

cation traces

magnesium and alkali salts (as sulfates): ≤3.0%

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium chloride (CaCl2) is inexpensive, easily available calcium salt with a wide range of applications. It is a key component in deicers as it melts snow and ice. Reports suggest that CaCl2 helps in preserving the firmness of fresh-cut fruits and vegetables during the storage.

Application

Calcium chloride was used during the assay of transglutaminase (TG) activity. It was also used for of modification of calcium alginate hydrogels.
It may be used in the following processes:
  • Modification of natural zeolite to increase its ability of fluoride removal.
  • Synthesis of calcium sulfate whiskers utilized in making polymer reinforcing composite materials.
  • Synthesis of calcium chloride-anodized aluminum composites, a solid sorbent.
  • As an extractant in the soil analysis process.
  • As a catalyst in the synthesis of aldols by reacting dimethylsilyl (DMS) enolates with aldehydes.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kehoe et al.
Journal of materials science. Materials in medicine, 22(4), 945-959 (2011-03-04)
The objective of this work was to examine the main (individual), combined (interaction) and second-order (quadratic) effects of: (i) poly(D,L-lactide-co-glycolide) (PLGA), (ii) F127, and (iii) a zinc-silicate based bioactive glass, on the cytotoxicity and ultimate tensile strength of an experimental
A review on the use of calcium chloride in applied thermal engineering.
N'Tsoukpoe KE, et al.
Applied Thermal Engineering, 75, 513-531 (2015)
Defluorination of wastewater by calcium chloride modified natural zeolite.
Zhang Z, et al.
Desalination, 276(1), 246-252 (2011)
Preparation of calcium chloride-anodized aluminum composite for water vapor sorption.
Kumita M, et al.
Applied Thermal Engineering, 50(2), 1564-1569 (2013)
Skin and soft tissue necrosis from calcium chloride in a deicer.
Kim MP, et al.
The Journal of Emergency Medicine, 32(1), 41-44 (2007)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service