Skip to Content
Merck
All Photos(2)

Documents

537454

Sigma-Aldrich

Butyl acetate

ReagentPlus®, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21

vapor density

4 (vs air)

Quality Level

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99.5%

form

liquid

autoignition temp.

790 °F

expl. lim.

7.6 %

impurities

≤0.05% (water)

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5.3 g/L)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl acetate (BA, n-butyl acetate) is an ester commonly used as an organic solvent in cosmetic and pharmaceutical industries and as a flavoring agent in food industry. One of the methods reported for its synthesis is the esterification reaction between acetic acid and butanol catalyzed by expandable graphite (EG). The decomposition of BA by photocatalysis in the gas phase has been studied using WO3/TiO2 (tungsten oxide/titanium dioxide), Pt/TiO2 and TiO2 catalysts. BA is a probable biofuel additive.

Application

Meets urethane grade requirements (H2O ≤ 0.05%)

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of trace gold in ores by butyl acetate extraction and flame atomic absorption spectrometry.
Shan XG and Lin YF.
Metallurgical Analysis, 3, 016-016 (2011)
Liquid-liquid equilibria for ternary systems acetic acid+ n-butyl acetate+ hydrocarbons at 293.15K.
Romain R, et al.
Fluid Phase Equilibria, 356, 264-270 (2013)
Catalytic Behaviors of Expansible Graphite in the Synthesis of Butyl Acetate.
Pang XY.
Journal of Chemistry, 9(4), 1816-1822 (2012)
Photocatalytic oxidation of butyl acetate in vapor phase on TiO2, Pt/TiO2 and WO3/TiO2 catalysts.
Keller V, et al.
J. Catal., 215(1), 129-138 (2003)
Ahmed Reda et al.
Frontiers in endocrinology, 5, 21-21 (2014-03-13)
Although three-dimensional testicular cell cultures have been demonstrated to mimic the organization of the testis in vivo and support spermatogenesis, the optimal culture conditions and requirements remain unknown. Therefore, utilizing an established three-dimensional cell culture system that promotes differentiation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service