Skip to Content
Merck
All Photos(2)

Key Documents

270687

Sigma-Aldrich

Butyl acetate

suitable for HPLC, 99.7%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
124-126 °C (lit.)
vapor pressure:
15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

grade

HPLC grade

Quality Level

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

Assay

99.7%

form

liquid

autoignition temp.

790 °F

purified by

glass distillation

expl. lim.

7.6 %

technique(s)

HPLC: suitable

impurities

≤0.05% water

evapn. residue

<0.0005%

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5.3 g/L)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 254 nm Amax: 1.0
λ: 260 nm Amax: 0.20
λ: 275 nm Amax: 0.04
λ: 300 nm Amax: 0.02
λ: 320-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fast Interfacial Defluorination Kinetics Enables Stable Cycling of Low-Temperature Lithium Metal Batteries.: Utilizing 2-Methyltetrahydrofuran as an electrolyte solvent, this research showcases how its unique properties contribute to the development of low-temperature lithium metal batteries with enhanced stability and efficiency, highlighting its critical role in advancing renewable energy technologies (Li et al., 2024).

  • α-Cleavage of 2,2-Diazido-2,3-dihydroinden-1-one in Solution and Cryogenic Matrices.: Demonstrating the use of 2-Methyltetrahydrofuran as a reaction medium, this study explores its effectiveness in facilitating controlled organic reactions under both solution and cryogenic conditions, providing insights into the mechanism of complex molecular transformations (James et al., 2024).

  • Solvent-Assisted Control of Metal-Organic Nanotube Size and Morphology.: This publication details the use of 2-Methyltetrahydrofuran in the synthesis of metal-organic frameworks, particularly focusing on its role in manipulating the size and shape of nanotubes, which are crucial for applications in catalysis and gas storage (Rosenmann et al., 2024).

  • Coordination Variations within Binuclear Copper Dioxygen-Derived (Hydro)Peroxo and Superoxo Species; Influences upon Thermodynamic and Electronic Properties.: Research utilizing 2-Methyltetrahydrofuran explores its application in complex inorganic chemistry, particularly in stabilizing reactive copper dioxygen complexes that are significant in biochemical oxygen transport and utilization (Hota et al., 2024).

  • Sustainable Green Extraction of Carotenoid Pigments: Innovative Technologies and Bio-Based Solvents.: This review highlights the role of 2-Methyltetrahydrofuran as a green solvent in the extraction of carotenoids, emphasizing its benefits in terms of efficiency and sustainability, and supporting the shift towards environmentally friendly extraction technologies (Morón-Ortiz et al., 2024).


Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pia Simona Bruni et al.
Forensic science international, 290, 103-110 (2018-07-18)
4-Hydroxy-N-methyl-N-ethyltryptamine (4-HO-MET) is a new psychoactive substance (NPS) of the chemical class of tryptamines. It shows structural similarities to the endogenous neurotransmitter serotonin, and is a serotonergic hallucinogen, affecting emotional, motoric, and cognitive functions. The knowledge about its biotransformation is
Limitations of radical thiol-ene reactions for polymer-polymer conjugation.
Koo SPS, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 48(8), 1699-1713 (2010)
Eagleson M.
Concise Encyclopedia Chemistry, 154-154 (1994)
Tim Goelen et al.
Pest management science, 77(2), 930-938 (2020-09-26)
Recent studies have shown that microorganisms emit volatile compounds that affect insect behaviour. However, it remains largely unclear whether microbes can be exploited as a source of attractants to improve biological control of insect pests. In this study, we used
The spectrophotometric and fluorometric determination of aluminum with 8-hydroxyquinoline and butyl acetate extraction.
Bloom PR, et al.
Soil Science Society of America Journal. Soil Science Society of America, 42(5), 713-716 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service