Skip to Content
Merck
All Photos(3)

Key Documents

443778

Sigma-Aldrich

NMP

ACS reagent, ≥99.0%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
202 °C (lit.)
81-82 °C/10 mmHg (lit.)
vapor pressure:
0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Product Name

1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%

grade

ACS reagent

Quality Level

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

dilution

(for analytical testing)

impurities

≤0.01% Free amines (as CH3NH2)
≤0.05% water

color

APHA: ≤50

refractive index

n20/D 1.47 (lit.)

pH

7.7-10.0 (20 °C, 100 g/L)

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

solubility

acetone: miscible(lit.)
alcohol: miscible(lit.)
benzene: miscible(lit.)
chloroform: miscible(lit.)
ethyl acetate: miscible(lit.)
water: miscible(lit.)

density

1.028 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methyl-2-pyrrolidinone may be used as:
  • An alternative eluent to THF in the size exclusion chromatography (SEC) based resolution of coal-derived liquids.
  • Reaction medium to prepare 2-aryl and 2-alkyl benzothiazoles.
  • A solvent in the ultrasonic preparation of metal organic framework (MOF-177) crystals.
It may be used to synthesize single-phase chalcopyrite CuInSe2 nanocrystals and potassium 3-methyl-2-oxo-pyrrolidinenitronate.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alkyl nitrate nitration of active methylene-compounds. 15. Nitration of 1-methyl-4-piperidone, 1-methyl-2-pyrrolidinone and 1,3-dimethyl-2-pyrrolidinone.
Feuer H, et al.
Journal of Heterocyclic Chemistry, 16(3), 481-485 (1979)
A Green and Air Pressure Solution Synthesis of CuInSe2 Nanocrystals Using 1-methyl-2-Pyrrolidinone as Hot Solvent.
Wang W, et al.
Journal of the American Ceramic Society. American Ceramic Society, 94(8), 2571-2577 (2011)
Aneuploidy induction in Saccharomyces cerevisiae by two solvent compounds, 1-methyl-2-pyrrolidinone and 2-pyrrolidinone.
Mayer VW, et al.
Environmental and Molecular Mutagenesis, 11(1), 31-40 (1988)
Solubility limitations in the determination of molecular mass distributions of coal liquefaction and hydrocracking products: 1-methyl-2-pyrrolidinone as mobile phase in size exclusion chromatography.
Herod AA, et al.
Energy and Fuels, 10(3), 743-750 (1996)
1-Methyl-2-pyrrolidinone: A well-adapted solvent in the benzothiazoles synthesis.
Brembilla A, et al.
Synthetic Communications, 20(21), 3379-3384 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service