Skip to Content
Merck
All Photos(2)

Key Documents

427144

Sigma-Aldrich

Poly(vinylidene fluoride)

average Mw ~275,000 by GPC, average Mn ~107,000, pellets

Synonym(s):

PVDF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CF2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

15 mmHg ( 32 °C)

form

pellets

melt index

7-20 g/10 min (230°C/12.5kg)

mol wt

average Mn ~107,000
average Mw ~275,000 by GPC

impact strength

2-4 ft-lb/in. (Izod, ASTM D 256, notched)

dielectric constant

8.0-9.5, 100 Hz (ASTM D 150)

hardness

76-80 (Shore D, ASTM D 2240)

refractive index

n20/D 1.42

ASTM D 3835

17,500-21,500 poise(230 °C) (100 sec-1)(lit.)

transition temp

brittleness temperature -62 °C (ASTM D 746)
Tm 166-170 °C (ASTM D 3418)

solubility

some polar solvents such as organic esters and amines: soluble

density

1.78 g/mL at 25 °C

SMILES string

FC(F)=C

InChI

1S/C2H2F2/c1-2(3)4/h1H2

InChI key

BQCIDUSAKPWEOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coatings, film, filter cloth, instrument linings, filtration membranes, pump parts and linings and valves.

Features and Benefits

Crystallizes in at least 3 forms, depending on crystallization conditions. High mechanical strength,abrasion resistance and dielectric strength. Resists most chemicals, UV radiation, weathering and fungi.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hanbit Jin et al.
ACS nano, 13(7), 7905-7912 (2019-06-28)
Soft and stretchable electrodes are essential components for skin-tight wearable devices, which can provide comfortable, unobtrusive, and accurate physiological monitoring and physical sensing for applications such as healthcare, medical treatment, and human-machine interfaces. Metal-elastomer nanocomposites are a promising approach, enabling
Lori Vidal Denham et al.
The Journal of the Acoustical Society of America, 132(3), 1368-1377 (2012-09-18)
Properties of a piezoelectric polymeric angioplasty balloon that may decrease the problems of acute closure and restenosis are evaluated in this study. Polyvinylidene difluoride (PVDF), a piezoelectric and pyroelectric polymer, has sufficient strength to serve as a standard angioplasty balloon
Eugene Cho et al.
Journal of nanoscience and nanotechnology, 13(1), 674-677 (2013-05-08)
Electrospun polyvinylidene fluoride (PVDF) nanofiber membranes have 3-dementional (3-D) open pore channel and hence have excellent application potential in Western blot. In this study we have modified electrospun PVDF nanofiber membrane by argon (Ar) plasma treatment to improve the surface
Noriko Kotobuki et al.
Journal of biomedical materials research. Part A, 101(2), 537-546 (2012-08-29)
For tissue engineering and regenerative medicine, stem cells should be effectively cultured in vitro. New thermoresponsive nanocomposite gels (MD-NC gels), consisting of inorganic clay (hectorite) and copolymers composed of hydrophobic 2-methoxyethyl acrylate (MEA) and hydrophilic N,N-dimethylacrylamide (DMAA) units, could be
Daisuke Matsutomo et al.
Journal of biochemistry, 153(2), 179-190 (2012-11-07)
Diacylglycerol kinase (DGK) participates in regulating the intracellular concentrations of two bioactive lipids, diacylglycerol and phosphatidic acid. DGKη1 is a type II isozyme that contains a pleckstrin homology (PH) domain and a pair of C1 domains at the N-terminus and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service