Skip to Content
Merck
All Photos(3)

Key Documents

360503

Sigma-Aldrich

2-Methoxyethanol

contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%

Synonym(s):

Ethylene glycol monomethyl ether, Methyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥99.3%
bp:
124-125 °C (lit.)
vapor pressure:
6.17 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

2.62 (vs air)

vapor pressure

6.17 mmHg ( 20 °C)

Assay

≥99.3%

form

liquid

autoignition temp.

548 °F

contains

50 ppm BHT as stabilizer

expl. lim.

24.5 %

impurities

≤0.002 meq/g Titr. acid
≤0.100% water

color

APHA: ≤10

refractive index

n20/D 1.402 (lit.)

pH

5.0-7.0 (25 °C)

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Ce-doped MgO films on AZ31 alloy substrate for biomedical applications: preparation, characterization and testing.: This study explores the preparation, characterization, and biomedical application of cerium-doped magnesium oxide films on AZ31 alloy substrates. The films were analyzed for their physical and chemical properties, and their potential use in biomedical devices was evaluated (Hattab et al., 2024).

  • A modified ionization difference UV-vis method for fast quantitation of guaiacyl-type phenolic hydroxyl groups in lignin.: This research presents a modified UV-vis spectroscopy method for the rapid quantification of guaiacyl-type phenolic hydroxyl groups in lignin. The study demonstrates improved accuracy and speed compared to traditional methods, making it valuable for lignin analysis in various applications (Chen et al., 2022).

  • Magnetism and spin dynamics of novel encapsulated iron oxide superparamagnetic nanoparticles.: This paper investigates the magnetic properties and spin dynamics of new encapsulated iron oxide superparamagnetic nanoparticles. The study provides insights into their potential applications in biomedical imaging and therapy (Arosio et al., 2013).

  • Anionic ring-opening polymerization of ethylene oxide in DMF with cyclodextrin derivatives as new initiators.: This study explores the use of cyclodextrin derivatives as initiators in the anionic ring-opening polymerization of ethylene oxide in DMF. The findings highlight the potential for developing new polymer materials with unique properties (Huin et al., 2013).

  • Synthesis of organosoluble chitosan derivatives with polyphenolic side chains.: The research focuses on the synthesis of organosoluble chitosan derivatives with polyphenolic side chains. The new derivatives exhibit improved solubility and potential for various industrial applications, including biomedical and environmental fields (Morimoto et al., 2012).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin Kirchberg et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 19(18), 2313-2320 (2018-06-07)
Mesoporous ZnFe2 O4 photoanodes have been prepared via dip-coating utilizing the evaporation-induced self-assembly of two different block-copolymer templates to investigate the influence of pore geometry on the photoelectrochemcial performance of those earth-abundant photoelectrodes. The use of commercial block copolymers, triblock
Conductometric, viscometric, and spectroscopic investigations on the solvation phenomena of alkali-metal ions and ion pairs in 2-methoxyethanol.
Das B and Hazra DK.
The Journal of Physical Chemistry, 99(1), 269-273 (1995)
Makoto Takei et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(2), 643-652 (2010-07-10)
Ethylene glycol monomethyl ether (EGME) is a widely used industrial solvent known to cause adverse effects to human and other mammals. Organs with high metabolism and rapid cell division, such as testes, are especially sensitive to its actions. In order
Anna Maria Piras et al.
International journal of pharmaceutics, 357(1-2), 260-271 (2008-03-04)
The preparation of novel biocompatible polymeric nanoconstructs suitable to load sensitive bioactive protein agents is reported. Nanoparticles were prepared as based on hybrid polymeric matrices consisting of synthetic bioerodible alternating copolymers of maleic anhydride and n-butylvinylether hemiesterified with 2-methoxyethanol and
H-Y Chang et al.
Occupational and environmental medicine, 61(8), 697-702 (2004-07-20)
To evaluate the protective effectiveness of gloves from occupational exposure to 2-methoxyethanol (2-ME); and to examine the association of 2-methoxyacetic acid (MAA) in urine and plasma collected simultaneously from low 2-ME exposure and high 2-ME exposure workers in a semiconductor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service