Skip to Content
Merck
All Photos(3)

Key Documents

338818

Sigma-Aldrich

Ammonium hydroxide solution

28% NH3 in H2O, ≥99.99% trace metals basis

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060103
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.99% trace metals basis

form

liquid

expl. lim.

27 %

concentration

28% NH3 in H2O

pH

11.7 (20 °C)

density

0.9 g/mL at 25 °C (lit.)
0.9 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium hydroxide solution is also called aqueous ammonia, aqua ammonia, or ammonia water. It is a weak base mainly used as a nitrogen source to prepare other nitrogen-containing organic and inorganic chemicals such as urea, ammonium carbonate, ammonium sulfate, etc.

Application

Ammonium hydroxide solution (28% NH3 in H2O) can be used as:
  • A medium to prepare magnetite (Fe3O4) nanoparticles via a modified controlled chemical coprecipitation method using divalent or trivalent iron salts as iron precursors.      
  • A reagent in ammonolysis.      
  • A solvent to synthesize 1, 4 dihydropyridines via one-pot condensation of an aldehyde and alkyl acetoacetate.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengqi Li et al.
Cell metabolism, 30(3), 508-524 (2019-06-18)
Fructose-1,6-bisphosphate (FBP) aldolase links sensing of declining glucose availability to AMPK activation via the lysosomal pathway. However, how aldolase transmits lack of occupancy by FBP to AMPK activation remains unclear. Here, we show that FBP-unoccupied aldolase interacts with and inhibits
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each
Aurélie Periat et al.
Journal of chromatography. A, 1356, 211-220 (2014-07-16)
In this study, the influence of electrospray ionization (ESI) source design on the overall sensitivity achieved in hydrophilic interaction chromatography (HILIC) and reversed phase liquid chromatography (RPLC), was investigated. State-of-the-art triple quadrupole mass analyzers from AB Sciex, Agilent Technologies and
J C Naviaux et al.
Translational psychiatry, 4, e400-e400 (2014-06-18)
Autism spectrum disorders (ASDs) now affect 1-2% of the children born in the United States. Hundreds of genetic, metabolic and environmental factors are known to increase the risk of ASD. Similar factors are known to influence the risk of schizophrenia
Mohammed Anwar et al.
International journal of pharmaceutics, 468(1-2), 158-164 (2014-04-22)
Nano-sized curcumin-loaded super-paramagnetic iron oxide nanoparticles (CUR-OA-SPIONs) were synthesized chemically by co-precipitation method using oleic acid as a stabilizer and Myrj 52 as a surfactant. The synthesized nanoparticles were characterized for their shape, size, surface morphology, electrokinetic potential, magnetic properties

Articles

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

See All

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service