Merck
  • Synthesis and in vitro localization study of curcumin-loaded SPIONs in a micro capillary for simulating a targeted drug delivery system.
Synthesis and in vitro localization study of curcumin-loaded SPIONs in a micro capillary for simulating a targeted drug delivery system.
International journal of pharmaceutics (2014-04-22)
Mohammed Anwar, Mohammed Asfer, Ayodhya P Prajapati, Sharmistha Mohapatra, Sohail Akhter, Asgar Ali, Farhan J Ahmad
ABSTRACT
Nano-sized curcumin-loaded super-paramagnetic iron oxide nanoparticles (CUR-OA-SPIONs) were synthesized chemically by co-precipitation method using oleic acid as a stabilizer and Myrj 52 as a surfactant. The synthesized nanoparticles were characterized for their shape, size, surface morphology, electrokinetic potential, magnetic properties, crystalinity, chemical interactions and thermal transitions. The synthesized CUR-OA-SPIONs were spherical, mono-dispersed, physically stable and super-paramagnetic in nature. In vitro localization study and aggregation dynamics of CUR-OA-SPIONs were studied with a flow of blood inside a square glass capillary (500×500 μm(2) cross section) in the presence of an externally applied magnetic field (Ms=1200 mT). This research which is first of its kind showed the fluorescent imaging of CUR-OA-SPIONs with respect to time to understand the aggregation dynamics of magnetic nanoparticles in a micro capillary simulating the case of targeted drug delivery system. The size of the aggregation increases with respect to time (t=0(+)s to t=500 s), while no significant change in the size of the aggregate was observed after time t=500 s.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Hexane, suitable for HPLC
Supelco
Curcumin, analytical standard
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Supelco
Hexane, analytical standard
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)