Skip to Content
Merck
All Photos(3)

Key Documents

270393

Sigma-Aldrich

Tetrachloroethylene

suitable for HPLC, ≥99.9%

Synonym(s):

PCE, Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CCl2
CAS Number:
Molecular Weight:
165.83
Beilstein:
1361721
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.04
Assay:
≥99.9%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
121 °C (lit.)
vapor pressure:
13 mmHg ( 20 °C)
19 mmHg ( 25 °C)

vapor density

5.83 (vs air)

Quality Level

vapor pressure

13 mmHg ( 20 °C)
19 mmHg ( 25 °C)

Assay

≥99.9%

form

liquid

purified by

glass distillation

technique(s)

HPLC: suitable

impurities

<0.050% water

evapn. residue

<0.0005%

color

APHA: ≤10

refractive index

n20/D 1.505 (lit.)

bp

121 °C (lit.)

mp

−22 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 290 nm Amax: 1.00
λ: 295 nm Amax: 0.30
λ: 300 nm Amax: ≤0.20
λ: 305 nm Amax: 0.10
λ: 350 nm Amax: 0.05
λ: 400 nm Amax: 0.03

application(s)

food and beverages

SMILES string

Cl\C(Cl)=C(\Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Dispersive liquid-liquid microextraction as a novel enrichment approach for compound-specific carbon isotope analysis of chlorinated phenols.: This study introduces a novel microextraction technique involving tetrachloroethylene, enhancing the sensitivity and specificity of carbon isotope analysis in complex environmental samples (Martin et al., 2024).

  • Photo-on-Demand In Situ One-Pot Synthesis of Carbonate Esters from Tetrachloroethylene.: This research presents a novel photochemical method for synthesizing carbonate esters from tetrachloroethylene, demonstrating its potential in synthetic organic chemistry and industrial applications (Higashimura et al., 2024).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dusty R V Berggren et al.
Environmental science & technology, 47(4), 1879-1886 (2013-01-16)
Results are presented from a chemostat study where the reductive dehalogenation of PCE was evaluated in the absence and presence of sulfate. Two chemostats inoculated with the Point Mugu culture, which contains strains of Dehalococcoides mccartyi, were operated at a
Annika S Fjordbøge et al.
Journal of contaminant hydrology, 140-141, 56-66 (2012-09-27)
Field investigations on the effects of ZVI-Clay soil mixing were conducted at a small DNAPL source zone with PCE as the parent compound. In a one-year monitoring program, soil samples were collected at three horizontal sampling planes (2.5, 5.0 and
Charline Wiegert et al.
Environmental science & technology, 46(20), 10918-10925 (2012-09-20)
Chlorinated ethenes (CEs) are ubiquitous groundwater contaminants, yet there remains a need for a method to efficiently monitor their in situ degradation. We report here the first field application of combined stable carbon and chlorine isotope analysis of tetrachloroethene (PCE)
Anika Reinhold et al.
Applied and environmental microbiology, 78(22), 8025-8032 (2012-09-11)
Corrinoids are essential cofactors of reductive dehalogenases in anaerobic bacteria. Microorganisms mediating reductive dechlorination as part of their energy metabolism are either capable of de novo corrinoid biosynthesis (e.g., Desulfitobacterium spp.) or dependent on exogenous vitamin B(12) (e.g., Dehalococcoides spp.).
Julie C Chambon et al.
Biotechnology and bioengineering, 110(1), 1-23 (2012-08-29)
Reductive dechlorination is a major degradation pathway of chlorinated ethenes in anaerobic subsurface environments, and reactive kinetic models describing the degradation process are needed in fate and transport models of these contaminants. However, reductive dechlorination is a complex biological process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service