Skip to Content
Merck
All Photos(1)

Key Documents

254312

Sigma-Aldrich

Titanium(IV) chloride

≥99.995% trace metals basis

Synonym(s):

TTC, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
liquid
liquid

grade

for analytical purposes

vapor pressure

50 mmHg ( 55 °C)
9.6 mmHg ( 20 °C)

Assay

≥99.995% trace metals basis

form

liquid
liquid

reaction suitability

reagent type: catalyst
core: titanium

bp

135-136 °C (lit.)
136.4 °C

mp

−25 °C (lit.)

density

1.73 g/mL at 20 °C (lit.)

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) chloride participates in the Baylis-Hillman reaction of arylaldehydes with methyl vinyl ketone. Neutral solutions of TiCl4 having a pH in the range of 2.5 to 6.0 (prepared by the addition of magnesium oxide as a base) are used to synthesize nanosized titanium dioxide.

Application

Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines.
Promotes a highly selective aldol reaction of (S)-2-benzyloxy-3-pentanone in THF or DME.
Titanium(IV) chloride has been used in the synthesis of anatase TiO2 structures with macropores (250nm in diameter) and mesoscale pores (50nm in diameter).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Review of Mesoporous TiO2 Thin Films
Wang J, et al.
Material Matters, 7(2) null
Victor Rodríguez-Cisterna et al.
The Journal of organic chemistry, 72(17), 6631-6633 (2007-07-20)
Stereoselectivity of TiCl4-mediated aldol reactions from (S)-2-benzyloxy-3-pentanone is dramatically improved when the reaction is carried out in the presence of 1.1 equiv of tetrahydrofuran (THF) or 1,2-dimethoxyethane (DME). The resultant 2,4-syn-4,5-syn adducts are then obtained in diastereomeric ratios up to
Precipitation of nanosized titanium dioxide from aqueous titanium (IV) chloride solutions by neutralization with MgO.
Li Y and Demopoulos GP.
Hydrometallurgy, 90(1), 26-33 (2008)
Titanium(IV) chloride and the amine-promoted baylis-hillman reaction
Shi et al.
Organic letters, 2(23), 3755-3755 (2000-11-14)
Bratindranath Mukherjee et al.
Nanoscale, 5(1), 269-274 (2012-11-20)
The surface treatment of an anodized TiO(2) nanotube (T_NT) is very desirable for enhancing its photoelectrochemical properties and often is a prerequisite to deposition of any overlying layer for photoactivity efficiency improvement. This study provides a comparative analysis of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service