Skip to Content
Merck
All Photos(1)

Key Documents

208566

Sigma-Aldrich

Titanium(IV) chloride

ReagentPlus®, 99.9% trace metals basis

Synonym(s):

TTC, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
Assay:
99.9% trace metals basis
form:
liquid

vapor pressure

50 mmHg ( 55 °C)
9.6 mmHg ( 20 °C)

Quality Level

100
200

product line

ReagentPlus®

Assay

99.9% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: titanium

bp

135-136 °C (lit.)

mp

−25 °C (lit.)

density

1.73 g/mL at 20 °C (lit.)

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) chloride participates in the Baylis-Hillman reaction of arylaldehydes with methyl vinyl ketone. Neutral solutions of TiCl4 having a pH in the range of 2.5 to 6.0 (prepared by the addition of magnesium oxide as a base) have been used for the synthesis of nanosized titanium dioxide.

Application

Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines.
Along with sodium borohydride, it forms an effective reagent for the reduction of:
  • carboxylic acid and acyl chloride to alcohols
  • carboxamides, oximes and lactams to amines
  • sulfoxides to sulfides
Titanium(IV) chloride (TiCl4) has been used as a precursor for the synthesis of Ag-TiO2@polyethylene glycol (PEG) nanoparticles. TiCl4 may also be used as one of the constituents to synthesize TiO2/Al2O3 binary oxide materials and N-(10-oxo-vinylanthracen-9-(10H)-ylidene)cyanamide.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaofei Xiao et al.
Journal of environmental sciences (China), 60, 14-23 (2017-10-17)
TiO
Reduction of some functional groups with titanium (IV) chloride/sodium borohydride.
Kano S, et al.
Synthesis, 1980(09), 695-697 (1980)
Scalable hybrid chemical manufacture to photothermal therapy: PEG-capped phototransducers
Byeon, Jeong Hoon
Scientific Reports, 6 (2016)
Influence of the sol-gel preparation method on the photocatalytic NO oxidation performance of TiO2/Al2O3 binary oxides.
Polat M, et al.
Catalysis Today, 241, 25-32 (2015)
Titanium(IV) chloride and the amine-promoted baylis-hillman reaction
Shi et al.
Organic letters, 2(23), 3755-3755 (2000-11-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service