Skip to Content
Merck
All Photos(5)

Key Documents

243418

Sigma-Aldrich

Potassium bromide

greener alternative

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050405
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
crystalline powder
crystalline solid
crystals or chunks

grade

ACS reagent

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

Assay

≥99.0%

form

crystalline powder
crystalline solid
crystals or chunks

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.005% insolubles

pH

5.0-8.8 (25 °C, 5%)

mp

734 °C (lit.)

anion traces

bromate (BrO3-): ≤0.001%
chloride (Cl-): ≤0.2%
iodate (IO3-): ≤0.001%
iodide (I-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
Mg: ≤0.001%
Na: ≤0.02%
heavy metals: ≤5 ppm (by ICP)

greener alternative category

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

May be used for removal of peripheral membrane proteins.

Other Notes

Go to Redi-Dri Product Listing 746444

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mild and regioselective oxidative bromination of anilines using potassium bromide and sodium perborate.
Roche D, et al.
Tetrahedron Letters, 41(33), 2083-2085 (2000)
Petr Suchomel et al.
PloS one, 10(3), e0119202-e0119202 (2015-03-18)
The diverse mechanism of antimicrobial activity of Ag and AgBr nanoparticles against gram-positive and gram-negative bacteria and also against several strains of candida was explored in this study. The AgBr nanoparticles (NPs) were prepared by simple precipitation of silver nitrate
Jacek Zielonka et al.
Free radical biology & medicine, 46(3), 329-338 (2008-11-26)
Methods for the detection and quantitation of hydroethidine (HE) and its oxidation products by HPLC analysis are described. Synthetic methods for preparation of authentic standards (2-hydroxyethidium and diethidium) are provided. Potential applications of the HPLC methods to chemical and biological
An efficient bromination of alkenes using cerium (IV) ammonium nitrate (CAN) and potassium bromide.
Nair V, et al.
Tetrahedron, 57(34), 7417-7422 (2001)
Density, viscosity, and electrolytic conductivity of concentrated aqueous electrolyte solutions at several temperatures. Alkaline-earth chlorides, lanthanum chloride, sodium chloride, sodium nitrate, sodium bromide, potassium nitrate, potassium bromide, and cadmium nitrate.
Isono T.
Journal of Chemical and Engineering Data, 29(1), 45-52 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service