Skip to Content
Merck
All Photos(1)

Key Documents

60089

Sigma-Aldrich

Potassium bromide

BioUltra, ≥99.5% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

Quality Level

product line

BioUltra

Assay

≥99.5% (AT)

form

solid

impurities

insoluble matter, passes filter test
≤0.001% total nitrogen (N)

loss

≤0.5% loss on drying, 110 °C

pH

5.0-7.0 (25 °C, 1 M in H2O)

mp

734 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

bromate (BrO3-): ≤10 mg/kg
chloride (Cl-): ≤1000 mg/kg
iodide (I-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium bromide is a chaotropic salt, mostly used as an anticonvulsant.

Application

Potassium bromide is used in the preparation of stable Pd nanocubes used in immunolabeling. Potassium bromide is used for the purification of Sarcocystis neurona sporocysts by discontinuous density gradient centrifugation.
May be used for removal of peripheral membrane proteins.

Biochem/physiol Actions

Potassium bromide is useful in the treatment of refractory seizures or tonic-clonic seizures. It executes a stable excitable membrane by neuron hyperpolarization.

Other Notes

In the resolution of flavoproteins into apoproteins and free flavins; Increases yield of extracellular SOD in the extraction from human tissues and cell lines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteome Analysis: Interpreting the Genome, 34-34 (2004)
M. Husain et al.
Methods in Enzymology, 53, 432-432 (1978)
Potassium bromide, an anticonvulsant, is effective at alleviating seizures in the Drosophila bang-sensitive mutant bang senseless
Tan JS, et al.
Brain Research, 1020(1-2), 45-52 (2004)
S L Marklund
The Journal of clinical investigation, 74(4), 1398-1403 (1984-10-01)
Extracellular-superoxide dismutase is a tetrameric copper-containing glycoprotein that previously has been demonstrated to be the major superoxide dismutase of human extracellular fluids. The occurrence of this enzyme in various human tissues that were removed from two accidental death victims and
Higson, S.P.J. et al.
Electroanalysis, 6, 431-431 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service