Skip to Content
Merck
All Photos(5)

Key Documents

217646

Sigma-Aldrich

Manganese(IV) oxide

activated, ~85%, <10 μm

Synonym(s):

Manganese dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

~85%

form

powder

particle size

<10 μm

mp

535 °C (dec.) (lit.)

SMILES string

O=[Mn]=O

InChI

1S/Mn.2O

InChI key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese (IV) oxide is a low cost, low toxic and easy handling inorganic compound widely used as a mild oxidant in organic chemistry. It is mainly used for the selective oxidation of activated alcohols to form α, β-unsaturated aldehydes and ketones.

Application

Manganese (IV) oxide can be used as selective oxidizing agent:
  • In conversion of allylic alcohols to α, β-ethylenic esters or amides.
  • In oxidation of allylic alcohols to α,β-ethylenic aldehydes or ketones.
  • In oxidation of propargylic alcohols, saturated alcohols and benzylic or heterocyclic alcohols.
  • Manganese (IV) oxide can also be used:,In hydration of nitriles to amides.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ross W Hogue et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(57), 14193-14199 (2017-06-09)
Self-assembly of 1:1:2 M
Mohammad Mahdi Najafpour
Dalton transactions (Cambridge, England : 2003), 40(15), 3805-3807 (2011-03-02)
A soluble form of colloidal manganese(IV) oxide showed efficient oxygen evolution or water oxidation in presence of oxone, H(2)O(2), cerium(IV) ammonium nitrate and tris(2,2'-bipyridyl)ruthenium(III).
Synthesis, characterization, and supercapacitor studies of manganese (IV) oxide nanowires.
Yousefi T, et al.
Materials Science in Semiconductor Processing, 16(3), 868-876 (2013)
Liling Jing et al.
Scientific reports, 10(1), 14937-14937 (2020-09-12)
Silver nanoparticles (Ag NPs) system capable of exhibiting different particle size at different temperature was developed, which depended on the extent of Diels-Alder (DA) reaction of bismaleimide with furan. Thus, Ag NPs were functionalized on the surface by a furyl-substituted
Water dispersible conducting nanocomposites of poly (N-vinylcarbazole), polypyrrole and polyaniline with nanodimensional manganese (IV) oxide.
Biswas M, et al.
Synthetic Metals, 105(2), 99-105 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service