918040
Manganese Oxide (MnO2) nanosheets
Synonym(s):
Birnessite manganese dioxide (δ-MnO2)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23
Recommended Products
form
liquid
Quality Level
application(s)
battery manufacturing
storage temp.
−20°C
SMILES string
O=[Mn]=O
InChI key
NUJOXMJBOLGQSY-UHFFFAOYSA-N
Related Categories
Application
Transition metal oxides (TMOs, eg, MnO2), have received a substantial amount of recent attention due to their distinct structure–property relationships in multiple fields, eg, optoelectronics, spintronics, catalysis, defect engineering, and energy-related applications. Birnessite manganese dioxide (δ-MnO2) nanosheets are highly regarded as an attractive electrode material for portable energy storage devices. As the layered structure of δ-MnO2 enables much thinner and flexible devices.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhong-Shuai Wu et al.
Advanced materials (Deerfield Beach, Fla.), 27(24), 3669-3675 (2015-05-15)
Ultrathin printable graphene supercapacitors are demonstrated, based on solution-processed electrochemically exfoliated graphene hybrid films on an ultrathin poly(ethylene terephthalate) substrate, exhibiting an unprecedented volumetric capacitance of 348 F cm(-3) , an ultrahigh scan rate of 2000 V s(-1) , and
Inkjet printing of δ-MnO2 nanosheets for flexible solid-state micro-supercapacitor
Wang Y, et al.
Nano Energy, 49, 481-488 (2018)
High-performance free-standing PEDOT:PSS electrodes for flexible and transparent all-solid-state supercapacitors.
Cheng T, et al.
Journal of Material Chemistry A, 4, 10493-10499 (2016)
Han Xiao et al.
ACS nano, 11(7), 7284-7292 (2017-06-20)
Rational engineering and simplified fabrication of high-energy micro-supercapacitors (MSCs) using graphene and other 2D nanosheets are of great value for flexible and integrated electronics. Here we develop one-step mask-assisted simplified fabrication of high-energy MSCs (PG-MSCs) based on the interdigital hybrid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service