Skip to Content
Merck
All Photos(3)

Key Documents

215074

Sigma-Aldrich

Potassium cyanate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOCN
CAS Number:
Molecular Weight:
81.12
Beilstein:
3594798
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38060513
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder, crystals or chunks
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

Assay

96%

form

powder, crystals or chunks

density

2.056 g/mL at 25 °C (lit.)

SMILES string

[K+].[O-]C#N

InChI

1S/CHNO.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

GKKCIDNWFBPDBW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium cyanate can be prepared by reacting urea with alkali hydroxide or carbonates at high temperatures.[1]

Application

Potassium cyanate may be used as one of the reaction components for the synthesis of the following:
  • urea[2]
  • hydantion[3]
  • glycosylamine 1,2-(cyclic carbamates) {glycofurano(or pyrano)[1,2-d]oxazolidin-2-ones}[4]
  • arylcarbamates[5]
  • (S)-(-)-1-carbamoyl-2-methoxymethylpyrrolidine[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S)-(-)-1-Amino-2-Methoxymethylpyrrolidine (SAMP) and (R)-(+)-1-Amino-2-Methoxymethylpyrrolidine (RAMP), Versatile Chiral Auxiliaries.
Enders D, et al
Organic Syntheses, 173-173 (1987)
Copper-Catalyzed Coupling of Arylboronic Acids with Potassium Cyanate: A New Approach to the Synthesis of Aryl Carbamates.
Kianmehr E and Baghersad MH.
Advanced Synthesis & Catalysis, 353(14-15), 2599-2603 (2011)
Transformation of aldoses into glycosylamine 1, 2-(cyclic carbamates)(glyco-oxazolidin-2-ones) by reaction with potassium cyanate.
Kovacs J, et al.
Carbohydrate Research, 210, 155-166 (1991)
Eagleson M.
Concise Encyclopedia Chemistry, 503-503 (1994)
Jeroen N Stoop et al.
PloS one, 9(7), e102163-e102163 (2014-07-16)
The immune response to post-translationally modified antigens is a key characteristic of rheumatoid arthritis. Carbamylation is such a posttranslational modification. Recently, we demonstrated that autoantibodies recognizing carbamylated proteins are present in sera of rheumatoid arthritis. The molecular mechanisms underlying the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service