Skip to Content
Merck
All Photos(3)

Key Documents

184497

Sigma-Aldrich

1,3-Dioxolane

ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%

Synonym(s):

Ethylene glycol methylene ether, Formaldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Beilstein:
102453
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
75-76 °C/1.013 hPa
vapor pressure:
70 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.6 (vs air)

Quality Level

vapor pressure

70 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

525 °F

contains

~75 ppm BHT as inhibitor

expl. lim.

2.1-20.5 %

refractive index

n20/D 1.401 (lit.)

bp

75-76 °C/1.013 hPa

mp

−95 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

C1COCO1

InChI

1S/C3H6O2/c1-2-5-3-4-1/h1-3H2

InChI key

WNXJIVFYUVYPPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dioxolane (DOXL) is a cyclic ether.[1] It is a green solvent.[2] It undergoes reaction with C60 to afford an epoxide and 1,3-dioxolane derivative. Reaction has been reported to proceed via a diradical mechanism.[3] A mixture of tetra(ethylene glycol) dimethyl ether (TEGDME) and DOXL has been used to compose the binary electrolyte for use in lithium-sulfur battery.[1] Its efficacy as a solvent in a non-aqueous redox flow battery system has been tested.[4]

Application

1,3-Dioxolane may be used in the fabrication of batteries and capacitors.[2] It may be used as one of the co-solvent to prepare the electrolyte for lithium-sulfur batteries.[5]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced electrochemical performance of a crosslinked polyaniline-coated graphene oxide-sulfur composite for rechargeable lithium-sulfur batteries.
Moon S, et al.
Journal of Power Sources, 294, 386-392 (2015)
Binary electrolyte based on tetra (ethylene glycol) dimethyl ether and 1, 3-dioxolane for lithium-sulfur battery.
Chang DR, et al.
Journal of Power Sources, 112(2), 452-460 (2002)
1, 3-Dioxolane: A green solvent for the preparation of carbon nanotube-modified electrodes.
Moscoso R, et al.
Electrochemical Communications, 48, 69-72 (2014)
Dong Zhou et al.
Nuclear medicine and biology, 35(6), 655-663 (2008-08-06)
Progesterone receptors (PRs) are present in many breast tumors, and their levels are increased by certain endocrine therapies. They can be used as targets for diagnostic imaging and radiotherapy. 16alpha,17alpha-[(R)-1'-alpha-(5-[(76)Br]Bromofurylmethylidene)dioxyl]-21-hydroxy-19-norpregn-4-ene-3,20-dione ([(76)Br]16alpha,17alpha-[(R)-1'-alpha-(5-bromofurylmethylidene)dioxyl]-21-hydroxy-19-norpregn-4-ene-3,20-dione (3)), a PR ligand with relative binding affinity (RBA)=65
Silvia Franchini et al.
European journal of medicinal chemistry, 45(9), 3740-3751 (2010-07-08)
A series of 1,3-dioxolane-based compounds incorporating a lactam (2-4) or imide (5-7) moiety was synthesized and the pharmacological profile at alpha(1)-adrenoceptor subtypes and 5-HT(1A) receptor was assessed through binding and functional experiments. Starting from the 2,2-diphenyl-1,3-dioxolane derivative 1, previously shown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service