Skip to Content
Merck
All Photos(2)

Key Documents

517925

Sigma-Aldrich

4′-Methylpropiophenone

90%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COC2H5
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

refractive index

n20/D 1.528 (lit.)

bp

238-239 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCC(=O)c1ccc(C)cc1

InChI

1S/C10H12O/c1-3-10(11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3

InChI key

PATYHUUYADUHQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4′-Methylpropiophenone can be used as a building block to synthesize:
  • N





  • -(4-Chlorophenyl)-N′-[1-(4-methylphenyl)propyl]urea as a potential CRAC channel inhibitor.
  • Mono and trinuclear cobaloxime/organocobaloxime complexes, which are used as efficient catalysts for the synthesis of cyclic carbonates.
  • Tolperisone (TOL) via acid-catalyzed reaction with piperidine hydrochloride and 1,2-dioxolane.

Other Notes

Remainder 3′-methylpropiophenone

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

V Karunakaran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 128, 1-14 (2014-03-25)
Combined experimental and theoretical studies have been performed on the structure and vibrational spectra (IR and Raman spectra) of 4'-methylpropiophenone (MPP). The FT-IR and FT-Raman spectra of 4'-methylpropiophenone (MPP) have been recorded in the region 4000-400 cm(-1) and 3500-100 cm(-1)
Alkaloid induced asymmetric electrocarboxylation of 4-methylpropiophenone.
Zhao SF, et al.
Tetrahedron Letters, 52(21), 2702-2705 (2011)
TLC-densitometric determination of tolperisone and its impurities 4-methylpropiophenone and piperidine in pharmaceutical preparations
Hubicka U, et al.
Journal of Liquid Chromatography and Related Technologies, 35(10), 1325-1335 (2012)
Ketone synthesized cobaloxime/organocobaloxime catalysts for cyclic carbonate synthesis from CO2 and epoxides: Characterization and electrochemistry
Kilic A, et al.
Journal of Organometallic Chemistry, 767, 150-159 (2014)
Paul I Dargan et al.
Drug testing and analysis, 3(7-8), 454-463 (2011-07-15)
Mephedrone (4-methylmethcathinone) is a synthetic cathinone that is used as a recreational drug. It has been available since 2007 but its availability and use increased significantly during 2009 and 2010. In this review article we will summarize the available literature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service