Skip to Content
Merck
All Photos(1)

Documents

06005

Sigma-Aldrich

Potassium hydroxide

puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1 mmHg ( 719 °C)

grade

puriss.

Assay

85-100.5%

form

pellets

quality

meets analytical specification of Ph. Eur., BP

impurities

residual solvents, complies
≤0.0005% heavy metals (as Pb)
≤0.0005% total nitrogen (N)
≤1% K2CO3

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
phosphate (PO43-): ≤10 mg/kg
silicate (as SiO2): ≤100 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤10 mg/kg
As: ≤3 mg/kg
Ca: ≤20 mg/kg
Fe: ≤10 mg/kg
Na: ≤1%

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide is an inorganic compound commonly known as caustic potash. It is frequently used to neutralize acids and prepare potassium salts of reagents. Additionally, NaOH is also employed in a variety of large-scale applications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuewang Huang et al.
Optics express, 23(5), 6780-6786 (2015-04-04)
We demonstrate a novel technique to fabricate sub-micron silicon nitride waveguides using conventional contact lithography with MEMS-grade photomasks. Potassium hydroxide anisotropic etching of silicon facilitates line reduction and roughness smoothing and is key to the technique. The fabricated waveguides is
Saeid Baroutian et al.
Bioresource technology, 102(2), 1095-1102 (2010-10-05)
In this study, a novel continuous reactor has been developed to produce high quality methyl esters (biodiesel) from palm oil. A microporous TiO2/Al2O3 membrane was packed with potassium hydroxide catalyst supported on palm shell activated carbon. The central composite design
Aldol condensation in gaseous phase by zeolite catalysts.
Wierzchowski PT and Zatorski LW.
Catalysis Letters, 9(5-6), 411-414 (1991)
Rokas Drevinskas et al.
Optics express, 23(2), 1428-1437 (2015-04-04)
Surface texturing is demonstrated by the combination of wet etching and ultrafast laser nanostructuring of silica glass. Using potassium hydroxide (KOH) at room temperature as an etchant of laser modified glass, we show the polarization dependent linear increase in retardance
Kinetics of palm oil transesterification in a batch reactor.
Darnoko D and Cheryan M.
Journal of the American Oil Chemists' Society, 77(12), 1263-1267 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service