Skip to Content
Merck
All Photos(1)

Key Documents

757551

Sigma-Aldrich

Potassium hydroxide

anhydrous, ≥99.95% trace metals basis

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.95% trace metals basis
grade:
anhydrous
form:
crystals or chunks
powder and chunks
powder or crystals
solubility:
H2O: soluble 1120 g/L
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 719 °C)

Assay

≥99.95% trace metals basis

form

crystals or chunks
powder and chunks
powder or crystals

impurities

≤500.0 ppm Trace Metal Analysis

mp

361 °C (lit.)

solubility

H2O: soluble 1120 g/L

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide (KOH) is also known as caustic potash. It is a strong alkali or a solid base compound that can be used in a variety of industrial applications. It finds its usage in the production of soaps, cleaners and detergents.[1]

Application

  • Biodiesel Production from Waste Cooking Oil: Describes the use of potassium hydroxide in biodiesel production, emphasizing its effectiveness in catalyzing the transesterification of waste cooking oil (K Komaladevi, R Pandithurai, 2023).
  • Chemical Activation for Dye Adsorption: Utilizes potassium hydroxide in the activation process to enhance the adsorption capacity of hemp-based activated carbon, crucial for environmental cleanup and management (Y Toptaş, B Yavuz, A Aksoğan Korkmaz, et al., 2024).
  • Surface Modification of Raphia Fibres: Highlights the use of potassium hydroxide for the chemical modification of Raphia fibres to improve their mechanical properties and water absorption, relevant in textile and material engineering (I Omemereyore, OT Uzoma, et al., 2024).
  • High-Performance Supercapacitors: Discusses the role of potassium hydroxide in enhancing the charge storage performance of electrodes, critical for the development of advanced energy storage systems (Y Pei, Z Ren, X Wu, et al., 2024).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The many uses of Potassium Compounds
Potassium (2007)
Mizuki Takemoto et al.
PloS one, 10(6), e0131094-e0131094 (2015-06-27)
Channelrhodopsin (ChR) is a light-gated cation channel that responds to blue light. Since ChR can be readily expressed in specific neurons to precisely control their activities by light, it has become a powerful tool in neuroscience. Although the recently solved
Ashok R Patel et al.
Journal of agricultural and food chemistry, 63(19), 4862-4869 (2015-05-02)
The aim of this study was to use a detailed rheological characterization to gain new insights into the gelation behavior of natural waxes. To make a comprehensive case, six natural waxes (differing in the relative proportion of chemical components: hydrocarbons
Nilesh M Agalave et al.
Pain, 155(9), 1802-1813 (2014-06-24)
Extracellular high mobility group box-1 protein (HMGB1) plays important roles in the pathogenesis of nerve injury- and cancer-induced pain. However, the involvement of spinal HMGB1 in arthritis-induced pain has not been examined previously and is the focus of this study.
Peng Li et al.
Angewandte Chemie (International ed. in English), 54(2), 574-577 (2014-11-15)
Self-assembly of a trigonal building subunit with diaminotriazines (DAT) functional groups leads to a unique rod-packing 3D microporous hydrogen-bonded organic framework (HOF-3). This material shows permanent porosity and demonstrates highly selective separation of C2H2/CO2 at ambient temperature and pressure.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service