Skip to Content
Merck
All Photos(1)

Documents

01148

Sigma-Aldrich

Sodium fluoride

puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050104
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1.4 mmHg ( 0 °C)

grade

puriss.

Assay

98.5-100.5% (calc. to the dried substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

acidity or alkalinity, complies
organic volatile impurities, complies (GC)
residual solvents, complies
≤0.002% heavy metals (as Pb)
≤0.1% free alkali (as NaOH)
≤0.2% free acid (as HF)
≤0.2% sodium hexafluorosilicate (Na2SiF6)

loss

≤0.5% loss on drying, 150 °C

mp

993 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Fe: ≤50 mg/kg

suitability

complies for appearance of solution

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium fluoride (NaF) is a metallic halide. It has been reported as dental caries preventive agent. It is the most widely employed preservative for glucose in blood. It has been reported to stabilize hepatic microsomal glucose-6-P phosphohydrolase enzyme.

Application

Sodium fluoride may be employed as fluorination agent in the synthesis of aromatic and aliphatic fluorides, via metathesis. It may be used to prepare the lysis buffer.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Drachman et al.
The Journal of biological chemistry, 270(10), 4979-4982 (1995-03-10)
c-Mpl is a member of the cytokine receptor superfamily, expressed primarily on hematopoietic cells. Recently, the c-Mpl ligand was cloned and found to have thrombopoietic activity. In this paper we report that ligand binding induced tyrosine phosphorylation in BaF3 cells
A J Lange et al.
The Journal of biological chemistry, 261(1), 101-107 (1986-01-05)
Stabilization and inhibition of hepatic microsomal glucose-6-P phosphohydrolase (EC 3.1.3.9) by F- requires the presence of Al3+ ions. At millimolar concentrations, reagent grade NaF inhibited glucose-6-P hydrolysis and protected the enzyme against inactivation induced by heat in the presence of
Effectiveness of sodium fluoride as a preservative of glucose in blood.
Chan AY, et al.
Clinical Chemistry, 35(2), 315-317 (1989)
Synthesis of fluorides by metathesis with sodium fluoride.
Tullock CW and Coffman DD.
The Journal of Organic Chemistry, 25(11), 2016-2019 (1960)
Hye-Jin Lee et al.
The Kaohsiung journal of medical sciences, 31(6), 303-308 (2015-06-05)
Dental caries preventive agents, such as sodium fluoride (NaF) and bamboo salt (BS), are known to cause cellular growth that is characterized by morphological and gene expression changes. This study was designed to investigate the dual effect of NaF and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service