Skip to Content
Merck
All Photos(1)

Key Documents

47072

Supelco

Fluoride ion solution for ISE

for ion-selective electrodes, 0.1 M F-, analytical standard (for ion-selective electrodes)

Synonym(s):

Sodium fluoride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
grade:
analytical standard (for ion-selective electrodes)
for ion-selective electrodes

grade

analytical standard (for ion-selective electrodes)
for ion-selective electrodes

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

0.1 M F-

density

1.005 g/mL at 25 °C (lit.)

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Preparation Note

prepared with NaF and H2O

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jocelyn Turpin et al.
Journal of virology, 89(2), 931-951 (2014-10-31)
Human T-cell lymphotropic virus type 1 (HTLV-1) and HTLV-2 encode auxiliary proteins that play important roles in viral replication, viral latency, and immune escape. The presence of auxiliary protein-encoding open reading frames (ORFs) in HTLV-3, the latest HTLV to be
Xianming Lin et al.
Channels (Austin, Tex.), 8(5), 433-443 (2014-12-09)
Connexin40 (Cx40) and connexin43 (Cx43) are co-expressed in the cardiovascular system, yet their ability to form functional heterotypic Cx43/Cx40 gap junctions remains controversial. We paired Cx43 or Cx40 stably-transfected N2a cells to examine the formation and biophysical properties of heterotypic
Bharat Jaishy et al.
Journal of lipid research, 56(3), 546-561 (2014-12-23)
Autophagy is a catabolic process involved in maintaining energy and organelle homeostasis. The relationship between obesity and the regulation of autophagy is cell type specific. Despite adverse consequences of obesity on cardiac structure and function, the contribution of altered cardiac
Irene Gallina et al.
Nature communications, 6, 6533-6533 (2015-03-31)
DNA replication stress is a source of genomic instability. Here we identify changed mutation rate 1 (Cmr1) as a factor involved in the response to DNA replication stress in Saccharomyces cerevisiae and show that Cmr1--together with Mrc1/Claspin, Pph3, the chaperonin
Jennifer D Lewis et al.
PloS one, 9(12), e116152-e116152 (2014-12-30)
Pseudomonas syringae employs a type III secretion system to inject 20-30 different type III effector (T3SE) proteins into plant host cells. A major role of T3SEs is to suppress plant immune responses and promote bacterial infection. The YopJ/HopZ acetyltransferases are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service