Skip to Content
Merck
All Photos(1)

Key Documents

Y0001580

Rocuronium bromide

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H53BrN2O4
CAS Number:
Molecular Weight:
609.68
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rocuronium

manufacturer/tradename

EDQM

solubility

H2O: 100 mg/mL, clear, light yellow

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N5CCOCC5)[N+]6(CCCC6)CC=C

InChI

1S/C32H53N2O4.BrH/c1-5-14-34(15-6-7-16-34)28-20-26-24-9-8-23-19-29(36)27(33-12-17-37-18-13-33)21-32(23,4)25(24)10-11-31(26,3)30(28)38-22(2)35;/h5,23-30,36H,1,6-21H2,2-4H3;1H/q+1;/p-1/t23-,24+,25-,26-,27-,28-,29-,30-,31-,32-;/m0./s1

InChI key

OYTJKRAYGYRUJK-FMCCZJBLSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rocuronium bromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Aminosteroid; neuromuscular blocker for anaesthesia

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Steinberg
Revista espanola de anestesiologia y reanimacion, 56(10), 604-611 (2010-02-16)
The correlation between gender and the potency of certain neuromuscular blockers, including rocuronium, has been investigated. However, the methods used have been inconsistent. This study aimed to look further at the role of gender in order to arrive at definitive
T Wang et al.
Anaesthesia, 69(8), 854-859 (2014-05-14)
We investigated whether laparoscopic vs open surgical approaches affected the duration of neuromuscular blockade following a single bolus dose of rocuronium. Fifty-three female patients underwent either laparoscopic or open gynaecological surgery. Rocuronium 0.6 mg.kg(-1) was administered to achieve neuromuscular blockade
J M Hunter
European journal of anaesthesiology. Supplement, 11, 39-41 (1995-09-01)
The pharmacokinetics of aminosteroid neuromuscular blocking drugs have been shown to be altered in patients with hepatic cirrhosis. A reduced clearance and prolonged elimination half-life of pancuronium and vecuronium have been demonstrated. Pharmacokinetic studies of the newest aminosteroid neuromuscular blocking
Hiroshi Iwasaki
Masui. The Japanese journal of anesthesiology, 55(7), 826-833 (2006-07-22)
For many years, much effort has been done to develop non-depolarizing neuromuscular blocking agents of rapid onset and short duration. It has been shown that rocuronium bromide is a nondepolarizing, steroidal muscle relaxant with fast onset and short-to-intermediate duration of
Facemask ventilation before or after neuromuscular blocking drugs: where are we now?
A Patel
Anaesthesia, 69(8), 811-815 (2014-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service