Skip to Content
Merck
All Photos(1)

Key Documents

Y0001208

Sinomenine

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO4
CAS Number:
Molecular Weight:
329.39
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sinomenine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C19H23NO4.ClH/c1-20-7-6-19-10-14(21)16(24-3)9-12(19)13(20)8-11-4-5-15(23-2)18(22)17(11)19;/h4-5,9,12-13,22H,6-8,10H2,1-3H3;1H/t12-,13+,19-;/m1./s1

InChI key

YMEVIMJAUHZFMW-VUIDNZEBSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sinomenine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Xu et al.
Planta medica, 74(12), 1423-1429 (2008-08-07)
Sinomenine (SIN), an alkaloid isolated from CAULIS SINOMENII, has been used in the treatment of rheumatoid arthritis (RA) clinically. This study aimed to systematically evaluate the efficacy and safety of SIN by a comparison between SIN and non-steroidal anti-inflammatory drugs
[Research advances of mechanism of sinomenine in treating rheumatoid arthritis].
Yong-Li Zhang et al.
Zhong xi yi jie he xue bao = Journal of Chinese integrative medicine, 7(8), 775-778 (2009-08-13)
Quanxing Wang et al.
International immunopharmacology, 11(3), 373-376 (2010-11-27)
Sinomenine (SN), a pure compound extracted from the Sinomenium acutum plant, has been found to inhibit T- and B-lymphocyte activation, proliferation and function and to interfere with the differentiation, recruitment and function of several other cell types, such as dendritic
Zhao Yang et al.
Molecular immunology, 60(2), 109-114 (2014-05-13)
Intracerebral hemorrhage (ICH) causes morbidity and mortality and commonly follows the reperfusion after an ischemic event. Microglial activation mediated cytokine and protease secretion contributes to brain injury in ICH. Previous studies have shown that sinomenine possesses potent immunoregulatory properties. However
Qing-Feng Kong et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 30(20), 1573-1576 (2006-01-21)
To further understand sinomenine, this paper has introduced the abstract technology, assaying, pharmaceutical chemistry, pharmacological action, pharmacotoxicology, pharmacokinetics and clinical application of sinomenine based on the important and significant contents of reference which have been consulted in the past ten

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service