Skip to Content
Merck
All Photos(1)

Key Documents

Y0001027

Betamethasone valerate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Betamethasone 17-valerate, 1,4-Pregnadiene-11β,17α,21-triol-9α-fluoro-16β-methyl-3,20-dione 17-valerate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17-valerate, 9α-Fluoro-16β-methylprednisolone 17-valerate, Betnovate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
Beilstein:
4240001
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCC(=O)O[C@@]1([C@@H](C)CC2C3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO

InChI

1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19?,20?,21-,24-,25-,26-,27-/m0/s1

InChI key

SNHRLVCMMWUAJD-QDHNOTTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone valerate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prakash Adhikari et al.
International journal of pediatric otorhinolaryngology, 75(4), 500-503 (2011-02-05)
This study was carried out with the objective of comparing clinical efficacy of 10% ichthammol glycerine (IG) pack with steroid-antibiotic pack for relieving pain in cases of acute otitis externa. A prospective quasi-randomized clinical trial was completely carried out in
Christine Bangert et al.
Dermatology (Basel, Switzerland), 222(1), 36-48 (2010-12-15)
Topical pimecrolimus may maintain remissions of atopic dermatitis (AD) by inhibiting subclinical inflammation. To evaluate clinical and cytological effects of pimecrolimus in topical corticosteroid-treated and resolved AD lesions. Patients (n=67) with resolved AD lesions were randomized to 3-week double-blind treatment
Atsuko Kamo et al.
Journal of dermatological science, 62(2), 91-97 (2011-04-05)
UV-based therapy has anti-pruritic effects in inflammatory skin diseases, such as atopic dermatitis and psoriasis. These anti-pruritic effects may be partly due to inhibition of intraepidermal nerve growth, but they have not been fully characterized. This study was performed to
Jens-Michael Jensen et al.
Experimental dermatology, 20(10), 783-788 (2011-06-29)
It has been suggested that the increased rate of bacterial infection in atopic dermatitis (AD) may be caused by reduced antimicrobial protein (AMP) expression. We were interested whether common treatments in AD affect antimicrobial defense. We investigated the effects of
Saif-ur-Rehman Khattak et al.
AAPS PharmSciTech, 14(1), 177-182 (2012-12-20)
The effects of solvent [acetonitrile, methanol, and acetonitrile/water mixture (20:80, v/v)], buffer concentration (phosphate buffer, pH 7.5), ionic strength and commonly employed adjuvants on the photodegradation of betamethasone-17 valerate in cream and gel formulations have been studied on exposure to UV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service