Skip to Content
Merck
All Photos(1)

Key Documents

B1000000

Betamethasone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9α-Fluoro-11β,17α,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16β-methylprednisolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

betamethasone

packaging

pkg of 60 mg

manufacturer/tradename

EDQM

storage condition

protect from light

color

white

mp

235-237 °C (lit.)
235.6 °C (456.1°F at 1013.0 hPa)

solubility

water: slightly soluble 0.063 g/L at 37 °C

density

0.305 g/cm3 at 25 °C (77°F)

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)[C@](O)(C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1

InChI key

UREBDLICKHMUKA-DVTGEIKXSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Betamethasone, a synthetic glucocorticoid, belongs to the corticosteroid family of active pharmaceutical ingredients (APIs).[1]

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone EP reference standard may be used as a pharmaceutical primary standard for the determination of the analyte in pharmaceutical formulations by chromatographic[1][2][3][4][5] and spectrophotometric techniques.[6][7]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Betamethasone
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 67(3), 4918-4920 (2021)
Analysis of betamethasone and its organic esters in pharmaceutical products
Graham RE, et al.
Journal of Pharmaceutical Sciences, 67(3), 360-363 (1978)
Simultaneous determination of dexamethasone and betamethasone in pharmaceuticals by reversed-phase HPLC
Santos-Montes A, et al.
Chromatographia, 39(9-10), 539-542 (1994)
Extraction-spectrophotometric method for the determination of betamethasone in pure form and in pharmaceutical formulations
Amin AS and Issa YM
Analytical Letters, 30(1), 69-78 (1997)
Elizabeth V Asztalos et al.
JAMA pediatrics, 167(12), 1102-1110 (2013-10-16)
A single course of antenatal corticosteroid therapy is recommended for pregnant women at risk of preterm birth between 24 and 33 weeks' gestational age. However, 50% of women remain pregnant 7 to 14 days later, leading to the question of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service