Skip to Content
Merck
All Photos(1)

Documents

Y0000867

Sertraline for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sertraline hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NCl2 · HCl
CAS Number:
Molecular Weight:
342.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sertraline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC1=C(Cl)C=CC([C@H]2C3=C(C=CC=C3)[C@@H](NC)CC2)=C1.[H]Cl

InChI

1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1

InChI key

BLFQGGGGFNSJKA-XHXSRVRCSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

Sertraline hydrochloride is a SSRI i.e selective serotonin reuptake inhibitor. It is mainly used in treating psychiatric disorders. It also is effective in treating hypotension occurring in disorders like neurocardiogenic syncope and idiopathic orthostatic hypotension which is affected by autonomic dysfunction.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sertraline for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective serotonin reuptake inhibitor, antidepressant.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yvette I Sheline et al.
The American journal of psychiatry, 169(11), 1185-1193 (2013-03-28)
In order to assess the effect of gray matter volumes and cortical thickness on antidepressant treatment response in late-life depression, the authors examined the relationship between brain regions identified a priori and Montgomery-Åsberg Depression Rating Scale (MADRS) scores over the
R Kaddurah-Daouk et al.
Translational psychiatry, 3, e223-e223 (2013-01-24)
In this study, we characterized early biochemical changes associated with sertraline and placebo administration and changes associated with a reduction in depressive symptoms in patients with major depressive disorder (MDD). MDD patients received sertraline or placebo in a double-blind 4-week
Quang A Le et al.
Health and quality of life outcomes, 11, 59-59 (2013-04-17)
To determine the minimal clinically important difference (MCID) for the health-utility measures EuroQol-5 dimensions (EQ-5D) and Quality of Well Being Self-Administered (QWB-SA) Scale in PTSD patients. Two hundred patients aged 18 to 65 years with PTSD enrolled in a doubly
Latif Moradveisi et al.
The British journal of psychiatry : the journal of mental science, 202(3), 204-211 (2013-02-09)
Behavioural activation might be a viable alternative to antidepressant medication for major depressive disorder. To compare the effectiveness of behavioural activation and treatment as usual (TAU, antidepressant medication) for major depressive disorder in routine clinical practice in Iran. Patients with
Alastair J Flint et al.
BMC psychiatry, 13, 38-38 (2013-01-29)
Psychotic depression (PD) is a severe disabling disorder with considerable morbidity and mortality. Electroconvulsive therapy and pharmacotherapy are each efficacious in the treatment of PD. Expert guidelines recommend the combination of antidepressant and antipsychotic medications in the acute pharmacologic treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service