Skip to Content
Merck
All Photos(1)

Key Documents

68317

Supelco

Ginsenoside Rg1

analytical standard

Synonym(s):

(3β,6α,12β)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-β-D-glucopyranoside, Ginsenoside A2, Ginsenoside g1, Panaxoside A, Panaxoside Rg1, Sanchinoside C1, Sanchinoside Rg1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@]12[C@@](C[C@H]([C@@]3([H])[C@]2(CC[C@]3([H])[C@@](CCC=C(C)C)(C)O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO)C)O)([H])[C@@]5([C@@](C(C)([C@H](CC5)O)C)([H])[C@H](C1)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6O)O)O)CO)C

InChI

1S/C42H72O14/c1-20(2)10-9-13-42(8,56-37-34(52)32(50)30(48)25(19-44)55-37)21-11-15-40(6)28(21)22(45)16-26-39(5)14-12-27(46)38(3,4)35(39)23(17-41(26,40)7)53-36-33(51)31(49)29(47)24(18-43)54-36/h10,21-37,43-52H,9,11-19H2,1-8H3/t21-,22+,23-,24+,25+,26+,27-,28-,29+,30+,31-,32-,33+,34+,35-,36+,37-,39+,40+,41+,42-/m0/s1

InChI key

YURJSTAIMNSZAE-HHNZYBFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginsenoside Rg1 is a major component of the root and stem of ginseng plant. It possesses a broad spectrum of pharmacological properties like neuroprotection, anti-inflammation, anti-aging, anti-fatigue and memory-enhancing properties.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuroprotective Effect of the Ginsenoside Rg1 on Cerebral Ischemic Injury In Vivo and In Vitro Is Mediated by PPAR?-Regulated Antioxidative and Anti-Inflammatory Pathways
Li Y, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM (2017)
Ginsenoside Rg1 protects against sepsis-associated encephalopathy through beclin 1?independent autophagy in mice
Li Y, et al.
The Journal of Surgical Research, 207, 181-189 (2017)
Wen-Fang Chen et al.
Menopause (New York, N.Y.), 19(9), 1052-1061 (2012-05-03)
The present study was designed to determine whether ginsenoside Rg1 could exert selective estrogenic effects by using both cell lines and an animal model. The endometrial Ishikawa cells and preosteoblastic MC3T3-E1 cells were treated with a different dose of Rg1.
X Zhang et al.
Neuroscience, 220, 191-200 (2012-06-26)
Ginsenoside Rg1, which could improve spatial learning and memory, might be a useful agent for preventing and treating cognitive impairment in Alzheimer's disease (AD). The present study was designed to test the neuroprotective effects of ginsenoside Rg1 on an ovariectomized
Yong Cheng et al.
Acta pharmacologica Sinica, 26(2), 143-149 (2005-01-25)
In the present paper, we overview the discovery of new biological activities induced by ginsenoside Rg1 and Rb1 and discuss possible mechanisms of action. Both compounds could increase neural plasticity in efficacy and structure; especially Rg1, as one small molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service