Skip to Content
Merck
All Photos(1)

Key Documents

Y0000543

Oxazepam for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Oxazepam, 7-Chloro-1,3-dihydro-3-hydroxy-5-phenyl-1,4(2H)-benzodiazepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClN2O2
CAS Number:
Molecular Weight:
286.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxazepam

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC1N=C(c2ccccc2)c3cc(Cl)ccc3NC1=O

InChI

1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-15(20)14(19)17-12/h1-8,15,20H,(H,17,19)

InChI key

ADIMAYPTOBDMTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxazepam for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anxiolytic; ligand for the GABAA receptor benzodiazepine modulatory site.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Lader
Acta psychiatrica Scandinavica. Supplementum, (274)(274), 89-93 (1978-01-01)
The benzodiazepines have been available now for about 15 years but new aspects of their actions are being elucidated all the time. Oxazepam has a short half-life and inactive metabolites and therefore differs from most other benzodiazepines. Some aspects of
Katarina Gospic et al.
PLoS biology, 9(5), e1001054-e1001054 (2011-05-12)
Imaging studies have revealed a putative neural account of emotional bias in decision making. However, it has been difficult in previous studies to identify the causal role of the different sub-regions involved in decision making. The Ultimatum Game (UG) is
Maarten Charles J Franck et al.
Patient education and counseling, 83(3), 411-416 (2011-05-31)
We developed a semi-automated leaflet optimizer (ABOP) to improve the readability of Dutch patient information leaflets (PILs). Our aim was to assess whether or not revisions made with ABOP have an effect on traceability and comprehension of PIL information. Two
Pharmacokinetics in clinical medicine: oxazepam versus other benzodiazepines.
D J Greenblatt et al.
Diseases of the nervous system, 36(5 Pt. 2), 6-13 (1975-05-01)
S A Lahousse et al.
Veterinary pathology, 48(4), 875-884 (2010-12-15)
Liver tumors from a previous National Toxicology Program study were examined using global gene expression and mutation analysis to define the mechanisms of carcinogenesis in mice exposed to oxazepam. Five hepatocellular adenomas and 5 hepatocellular carcinomas from male B6C3F1 mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service