Skip to Content
Merck
All Photos(1)

Key Documents

Y0000458

Repaglinide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Repaglinide, (S)-(+)-2-Ethoxy-4-[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]aminocarbonylmethyl]benzoic acid, Novonorm, Prandin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36N2O4
CAS Number:
Molecular Weight:
452.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

repaglinide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOc1cc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)ccc1C(O)=O

InChI

1S/C27H36N2O4/c1-4-33-25-17-20(12-13-22(25)27(31)32)18-26(30)28-23(16-19(2)3)21-10-6-7-11-24(21)29-14-8-5-9-15-29/h6-7,10-13,17,19,23H,4-5,8-9,14-16,18H2,1-3H3,(H,28,30)(H,31,32)/t23-/m0/s1

InChI key

FAEKWTJYAYMJKF-QHCPKHFHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Repaglinide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Repaglinide is a potent short-acting insulin secretagogue that acts by closing ATP-sensitive potassium (KATP) channels in the plasma membrane of the pancreatic beta cell. It represents a new class of insulin secretagogues, structurally unrelated to sulphonylureas, which were developed for the treatment of type 2 diabetes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Xiang et al.
Pharmacology, 89(1-2), 105-110 (2012-03-09)
The aim of this study was to evaluate the pharmacogenetic variability in the disposition of repaglinide in healthy Chinese subjects. A single dose of 2 mg repaglinide was orally administered to 24 healthy Chinese subjects. The serum concentrations of repaglinide
Christina Jarnert et al.
Diabetes & vascular disease research, 9(4), 287-295 (2012-03-02)
Microcirculatory and endothelial dysfunction are signs of cardiovascular engagement in patients with type 2 diabetes. This study tested whether glucose normalisation may reverse this. Thirty-nine T2DM patients (age 61±7 years, 58% females) with signs of mild diastolic dysfunction were randomised
Deaths in trials should always be reported.
Jeppe Schroll
BMJ (Clinical research ed.), 347, f4219-f4219 (2013-07-06)
Kuldeep Sharma et al.
Biomedical chromatography : BMC, 27(3), 356-364 (2012-08-07)
A highly sensitive and specific LC-MS/MS-ESI method has been developed for simultaneous quantification of metformin (MFN) and repaglinide (RGN) in rat plasma (50 μL) using phenacetin as an internal standard (IS). Simple protein precipitation was used to extract MFN and
Boggarapu Prakash Rao et al.
Pakistan journal of pharmaceutical sciences, 26(4), 707-713 (2013-07-03)
The full factorial design was employed to evaluate contribution of drug: polymer and Eudragit RS 100: Eudragit RL 100 on entrapment efficiency, time for maximum drug release, percentage of drug release. Floating microparticles were prepared using spray drying technique. Microparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service