Skip to Content
Merck
All Photos(1)

Key Documents

Y0000433

Ammonium glycyrrhizate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glycyrrhizic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H62O16 · NH3
CAS Number:
Molecular Weight:
839.96
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glycyrrhizic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(O)=O)O[C@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)C(O)=O)C(O)=O

InChI

1S/C42H62O16.H3N/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);1H3/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+;/m0./s1

InChI key

ILRKKHJEINIICQ-OOFFSTKBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ammonium glycyrrhizate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Okuma et al.
Neuropharmacology, 85, 18-26 (2014-05-27)
Glycyrrhizin (GL) is a major constituent of licorice root and has been suggested to inhibit the release of high mobility group box-1 (HMGB1), a protein considered representative of damage-associated molecular patterns. We found that GL bound HMGB1 but not RAGE
Yusuke Izutani et al.
Carbohydrate research, 392, 25-30 (2014-05-23)
The structural properties of glycyrrhizic acid, a sweet-tasting constituent of Glycyrrhiza glabra, and its interaction with cyclodextrins were analyzed using dynamic light scattering, isothermal titration calorimetry, and NMR. The dynamic light scattering and NMR studies showed that glycyrrhizic acid forms
Chiung-Hung Chang et al.
PloS one, 9(8), e105362-e105362 (2014-08-19)
Salmonella, a common zoonotic pathogen, causes gastroenteritis in both humans and animals. Traditional Chinese Medicine (TCM) has been used to improve gastrointestinal dysfunction and to modify the immune response to inflammation for centuries. This study used six herbal plants and
Pil-Sung Yang et al.
Respiratory research, 15, 148-148 (2014-11-26)
High mobility group box-1 (HMGB1), a proinflammatory cytokine, plays a pivotal role in tissue remodeling and angiogenesis, both of which are crucial for the pathogenesis of pulmonary arterial hypertension. In this study, we explored the relationship between HMGB1 and pulmonary
Hye Young Kim et al.
PloS one, 9(8), e104743-e104743 (2014-08-16)
High mobility group box chromosomal protein 1 (HMGB-1) released from injured cells plays an important role in the development of arthritis. This study investigated the anti-angiogenic effects of cilostazol in collagen-induced arthritis (CIA) of mice, and the underlying mechanisms involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service