Skip to Content
Merck
All Photos(3)

Key Documents

T4062

Sigma-Aldrich

Tetracycline hydrochloride

meets USP testing specifications

Synonym(s):

Tetracycline HCL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Beilstein:
3844873
EC Number:
MDL number:
UNSPSC Code:
51284043
PubChem Substance ID:
NACRES:
NA.76

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

≥900 (Units in µg/mg)

form

crystalline powder

optical activity

[α]/D -255 to 240° (Specific rotation )

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Light sensitive.)

impurities

≤4.0%

color

yellow

mp

220-223 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1

InChI key

XMEVHPAGJVLHIG-FMZCEJRJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Tetracycline is a broad spectrum polyketide antibiotic with clinical uses in treating bacterial infections such as Rocky Mountain spotted fever, typush fever, tick fevers, Q fever, and Brill-Zinsser disease and to treat upper respiratory infections and acne. It has been used in studies of multidrug resistance and potential side effects including acute pancreatitis. It is recommended for use in cell culture applications at 10 mg/L.

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.

Caution

This product should be frozen below 0°C and protected from light and moisture. In these conditions, the product has been shown to retain activity for 4 years. Stock solutions should be stored at -20°C and are stable at 37°C for 4 days.

Preparation Note

The product is freely soluble in water, soluble in methanol and ethanol but is insoluble in ether and hydrocarbons. In water, the product yields a clear, yellow-orange solution with heating and in 95% ethanol, 50 mg dissolved in 4 mL with heating yields a clear, yellow-green solution. Tetracycline is rapidly destroyed by alkali hydroxide solutions and standing water solutions become turbid due to hydrolysis and precipitation. The potency of tetracycline is reduced in solutions with pH below 2.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Light sensitive.

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shruti Sharma et al.
Nature communications, 10(1), 3105-3105 (2019-07-17)
Fas plays a major role in regulating ligand-induced apoptosis in many cell types. It is well known that several cancers demonstrate reduced cell surface levels of Fas and thus escape a potential control system via ligand-induced apoptosis, although underlying mechanisms
Yasuhiro Igarashi et al.
Journal of natural products, 74(4), 670-674 (2011-03-11)
A new spirotetronate-class polyketide, maklamicin (1), was isolated from the culture extract of an endophytic actinomycete of the genus Micromonospora. The structure and relative configuration of 1 were elucidated by interpretation of NMR and other spectroscopic data, and the absolute
Ruiling Qi et al.
Colloids and surfaces. B, Biointerfaces, 110, 148-155 (2013-05-29)
Fabrication of nanofiber-based drug delivery system with controlled release property is of general interest in biomedical sciences. In this study, we prepared an antibiotic drug tetracycline hydrochloride (TCH)-loaded halloysite nanotubes/poly(lactic-co-glycolic acid) composite nanofibers (TCH/HNTs/PLGA), and evaluated the drug release and
Zhijie Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(13), 5004-5009 (2013-03-12)
Reported here is a piggyBac transposon-based expression system for the generation of doxycycline-inducible, stably transfected mammalian cell cultures for large-scale protein production. The system works with commonly used adherent and suspension-adapted mammalian cell lines and requires only a single transfection
T Bouchery et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 19(2), 131-140 (2013-02-13)
Symbiotic associations between eukaryotes and microorganisms are frequently observed in nature, and range along the continuum between parasitism and mutualism. The genus Wolbachia contains well-known intracellular bacteria of arthropods that induce several reproductive phenotypes that benefit the transmission of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service