Skip to Content
Merck
All Photos(1)

Documents

46133

Supelco

Chlortetracycline hydrochloride

VETRANAL®, analytical standard

Synonym(s):

7-Chlorotetracycline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23ClN2O8 · HCl
CAS Number:
Molecular Weight:
515.34
Beilstein:
3858364
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E702
NACRES:
NA.24

grade

analytical standard

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

210-215 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

cleaning products
clinical
cosmetics
environmental
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2CC3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O

InChI

1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7?,8-,15-,21-,22-;/m0./s1

InChI key

CBHYYLPALVVVEY-LYNLVHCPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luxi Dong et al.
Chemosphere, 89(1), 44-51 (2012-06-01)
DNA damage and changes in enzyme activities were used as biomarkers to evaluate the genotoxicity and oxidative stress of tetracycline and chlortetracycline on the earthworm Eisenia fetida. The results showed that both antibiotics induced significant genotoxicity on earthworms in a
Hui Fu et al.
Wei sheng yan jiu = Journal of hygiene research, 41(2), 282-286 (2012-05-23)
To develop a method for determination of the Metronidazole, Chlortetracycline hydrochloride, Oxytetracycline dihydrate, Minocycline hydrochloride, Erythromycin and Tetracycline hydrochloride in disinfection products by high performance liquid chromatography tandem mass spectrometry(LC-MS/MS). Samples were extracted by methanol and filtered through 0.45 microm
Hyeon Young Kim et al.
Bulletin of environmental contamination and toxicology, 89(2), 234-239 (2012-06-08)
The purpose of this study was to examine the effects of veterinary antibiotics, including amoxicillin (AMX), chlortetracycline (CTC) and tylosin (TYL), on the biochemical mechanism of human embryonic kidney cells (HEK293). CTC and TYL inhibited HEK293 cell proliferation, in both
V V Shepelevich et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 74(6), 24-28 (2013-01-09)
Examination of sensitivity of 10 Pseudomonas chlororaphis strains belonging to different subspecies to 54 antibiotics has shown that all studied representatives of Pchlororaphis subsp. chlororaphis, P. chlororaphis subsp. aureofaciens and Pchlororaphis subsp. aurantiaca were sensitive to aminoglycoside antibiotics and fluoroquinolones
Dasong Lin et al.
Environmental pollution (Barking, Essex : 1987), 171, 46-51 (2012-08-08)
This study aims to evaluate toxic effects of exposure to chlortetracycline (CTC) in soil on reproductive endpoints (juvenile counts and cocoon counts), biochemical responses, and genotoxic potentials of the earthworm Eisenia fetida. Results showed that juvenile counts and cocoon counts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service