Skip to Content
Merck
All Photos(1)

Key Documents

STS0005

Sigma-Aldrich

Triton CG-110

greener alternative

Non-ionic surfactant

Synonym(s):

Alkyl polyglucoside (APG)-natural surfactant

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.21

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

1748 ppm (25°C)

transition temp

pour point -15 °C
cloud point >100 °C (1 wt% actives aq solution)

solubility

water: freely soluble at 20 °C (visual)

density

1.15 g/mL at 20 °C

greener alternative category

SMILES string

O1[C@@H]([C@H]([C@@H]([C@H](C1OCCCCCCCCCC)O)O)O)CO

InChI

1S/C16H32O6/c1-2-3-4-5-6-7-8-9-10-21-16-15(20)14(19)13(18)12(11-17)22-16/h12-20H,2-11H2,1H3/t12-,13-,14+,15-,16?/m1/s1

InChI key

JDRSMPFHFNXQRB-IWQYDBTJSA-N

General description

Triton CG-110 is a mixture of 58.0-62.0% D-glucopyranose, oligomeric, decyl octyl glycoside and 38.0-42.0% water. Contains less than 2% decanol and less than 1.0% octanol.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Application

Triton CG-110 can be used:
  • In the preparation of multilamellar vesicles as carriers for tretinoins and retinoids.
  • In the study of mineralization, leaching, and adsorption of glyphosate herbicide in the soil.

Features and Benefits

  • Mild
  • Good detergency & wetting properties
  • Lower color than Triton BG-10
  • Readily biodegradable
  • Moderate to high stable foam

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Carretta et al.
Journal of hazardous materials, 388, 122111-122111 (2020-01-21)
Glyphosate is the most common herbicide worldwide, and its impact on the environment has increasingly been scrutinized. Glyphosate-based formulations can contain co-formulants, among which are surfactants. This study aimed to investigate whether the presence of an alkyl polyglucoside-based surfactant, Triton
Niosomes as carriers for tretinoin. I. Preparation and properties
Manconi M, et al.
International Journal of Pharmaceutics, 234(1-2), 237-248 (2002)
Decyl glucoside surfactant Triton CG-110 does not significantly affect the environmental fate of glyphosate in the soil at environmentally relevant concentrations
Carretta L, et al.
Journal of Hazardous Materials, 6(5), 122111-122111 (2020)
Retinoids: new use by innovative drug-delivery systems
Trapasso E, et al.
Expert Opinion on Drug Delivery, 6(5), 465-483 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service