Skip to Content
Merck
All Photos(1)

Key Documents

769320

Sigma-Aldrich

Sodium azide

≥99%, ultra dry

Synonym(s):

Azide, Hydrazoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN3
CAS Number:
Molecular Weight:
65.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

Assay

≥99%
99.0-101.0% (RT)

form

solid

quality

ultra dry

impurities

≤0.05% insolubles in water
≤0.2% free alkaline (as NaOH)

loss

≤0.2% loss on drying

cation traces

heavy metals (as Pb): ≤0.001%

SMILES string

[Na]N=[N+]=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium azide is well known for its application in automobile air bags. It also find use in organic synthesis involving azide functionalization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takatoshi Nohara et al.
Journal of biomaterials science. Polymer edition, 28(10-12), 925-938 (2017-04-21)
The in vitro enzymatic synthesis of cellulose and its derivatives has attracted growing interest for fabricating novel cellulose-based supramolecular assemblies with unique physicochemical and functional properties. However, their potential biomedical applications have not been sufficiently demonstrated despite their useful features
Malinda Salim et al.
Molecular pharmaceutics, 15(8), 3535-3544 (2018-06-23)
Milk has been used as a vehicle for the delivery of antimalarial drugs during clinical trials to test for a food effect and artefenomel (OZ439) showed enhanced oral bioavailability with milk. However, the nature of the interaction between milk and
Reductive azidation of carbonyl compounds via tosylhydrazone intermediates using sodium azide.
Barluenga, J., et al.
Angewandte Chemie (International Edition in English), 51, 5950-5952 (2012)
Matthias Rieckher et al.
Cell reports, 25(1), 199-211 (2018-10-04)
Aging is accompanied by a pervasive collapse of proteostasis, while reducing general protein synthesis promotes longevity across taxa. Here, we show that the eIF4E isoform IFE-2 is increasingly sequestered in mRNA processing (P) bodies during aging and upon stress in
Eunbyeol Seo et al.
Scientific reports, 9(1), 18648-18648 (2019-12-11)
Chromic dyes responding against external stimuli are useful in various field of applications especially to colorimetric sensors. However, there have been several limitations in generic application because of its cost, stability and reliability. Here, we introduced highly functionalizable polymeric materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service