Skip to Content
Merck
All Photos(1)

Key Documents

PHR2408

Supelco

N-Nitrosodiethylamine (NDEA)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

N-Nitrosodiethylamine, Diethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NNO
CAS Number:
Molecular Weight:
102.14
Beilstein:
1744991
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1466652

form

liquid

CofA

current certificate can be downloaded

packaging

pkg of 1 g

bp

177 °C (lit.)

density

0.95 g/mL (lit.)

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

CCN(CC)N=O

InChI

1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3

InChI key

WBNQDOYYEUMPFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

N-Nitrosodiethylamine (NDEA) pharmaceutical secondary referencestandard is used to determine NDEA impurity in:
  • Lisinopril API and its final product
  • Sitagliptin and metformin hydrochlorideimmediate and extended-release formulations
These Secondary Standards are qualified as Certified ReferenceMaterials. These are suitable for use in several analytical applicationsincluding but not limited to pharma release testing, pharma method developmentfor qualitative and quantitative analyses, food and beverage quality controltesting, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4356 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of an HPLC-FLD Method for the Determination of NDMA and NDEA Nitrosamines in Lisinopril Using Pre-Column Denitrosation and Derivatization Procedure
Tsanaktsidou, et al
Separations, 9, 347-347 (2022)
Development and validation of rapid ultra high performance liquid chromatography with tandem mass spectroscopic method for the quantification of N-nitrosodimethyl amine and N-nitrosodiethyl amine in sitagliptin and metformin hydrochloride immediate and extended-release formulations
Mallavarapu, et al.
Journal of Separation Science, 45, 3067-3081 (2022)

Articles

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

See All

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service