Skip to Content
Merck
All Photos(1)

Key Documents

PHR1235

Supelco

Retinyl Palmitate (Vitamin A Palmitate)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Retinyl palmitate, Vitamin A palmitate, all−trans−Retinol palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O2
CAS Number:
Molecular Weight:
524.86
Beilstein:
1917366
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1602502

API family

retinyl compounds

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/COC(CCCCCCCCCCCCCCC)=O)C(C)(C)CCC1

InChI

1S/C36H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-25-35(37)38-30-28-32(3)23-20-22-31(2)26-27-34-33(4)24-21-29-36(34,5)6/h20,22-23,26-28H,7-19,21,24-25,29-30H2,1-6H3/b23-20+,27-26+,31-22+,32-28+

InChI key

VYGQUTWHTHXGQB-FFHKNEKCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.Retinyl palmitate belongs to a category of compounds called retinoids, which are chemically similar to vitamin A. It exhibits a beneficial effect on vision, skin and immune function, inhibits cell proliferation and prevents cancer. It is an important dietary as well as a therapeutic compound.

Application

Retinyl palmitate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatographic and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Review: Vitamin A metabolism.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1694 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a method for quantitation of retinol and retinyl palmitate in human serum using high-performance liquid chromatography -atmospheric pressure chemical ionization -mass spectrometry
van Breemen RB, et al.
Journal of Chromatography A, 794(1-2), 245-251 (1998)
Vitamin A Palmitate-β-cyclodextrin inclusion complexes: Characterization, protection and emulsification properties
Vilanova N and Solans C
Food Chemistry, 175(1-2), 529 -5535 (2015)
S Pedersen-Bjergaard et al.
Journal of chromatography. A, 876(1-2), 201-211 (2000-05-24)
Microemulsion electrokinetic chromatography (MEEKC) was carried out in a pH 2.5 phosphate buffer to effectively suppress the electroosmotic flow (EOF). With 66.6% (w/w) 25 mM phosphate buffer pH 2.5, 20.0% (w/w) 2-propanol, 6.6% (w/w) 1-butanol, 6.0% (w/w) sodium lauryl sulphate
T W de Bruin et al.
The American journal of clinical nutrition, 58(4), 477-483 (1993-10-01)
The postprandial lipoprotein metabolism of two orally administered vitamin A-fat loads consisting of either 20% (wt:vol) soybean oil or 17% olive oil plus 3% soybean oil was studied in six normolipidemic young men according to a randomized crossover design. Mean
G Tzimas et al.
The Journal of nutrition, 126(9), 2159-2171 (1996-09-01)
Retinoid pharmacokinetics were examined in plasma, placenta and embryos of gestational d 12 rabbits following application of an embryotoxic dosing regimen (10 mg retinyl palmitate/kg body wt per day from gestational d 7 to 12). Vehicle-treated or untreated rabbits served

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service