Skip to Content
Merck
All Photos(2)

Key Documents

P6585

Sigma-Aldrich

TWEEN® 20

Low-peroxide; Low-carbonyls

Synonym(s):

Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

Quality Level

description

non-ionic

form

viscous liquid

mol wt

~1228

contains

BHT as antioxidant

impurities

≤0.5 μmol/g Peroxides
≤1.0 μmol/g Carbonyls
≤3.0% water

refractive index

n20/D 1.468 (lit.)

CMC

0.06 mM (20-25°C)

transition temp

cloud point 76 °C

density

1.095 g/mL at 25 °C (lit.)
1.1 g/cm3 at 25 °C (lit.)

HLB

16.7

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tween®20 is a polyoxyethylene sorbitol ester that belongs to the polysorbate family. It is a nonionic detergent having a molecular weight of 1,225 daltons, assuming 20 ethylene oxide units, 1 sorbitol, and 1 lauric acid as the primary fatty acid. The ethylene oxide subunits are responsible for the hydrophilic nature of the surfactant, while the hydrocarbon chains provide the hydrophobic environment. Sorbitol forms the backbone ring to which the ethylene oxide polymers are attached.

Application

Non-ionic detergent
Tween 20 has been used along with PBS to wash plates in ELISA technique.

Preparation Note

Purified to remove peroxides and aldehydes

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

527.0 °F - Pensky-Martens closed cup

Flash Point(C)

275 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poke but don't pinch: risk assessment and venom metering in the western black widow spider, Latrodectus hesperus.
Nelsen DR
Animal behaviour, 89, 107-114 (2014)
Polysorbates 20 and 80 used in the formulation of protein biotherapeutics: structure and degradation pathways.
Kerwin BA
Journal of Pharmaceutical Sciences, 97(8), 2924-2935 (2008)
John Ong et al.
Scientific reports, 10(1), 12429-12429 (2020-07-28)
Albumin-based hydrogels are increasingly attractive in tissue engineering because they provide a xeno-free, biocompatible and potentially patient-specific platform for tissue engineering and drug delivery. The majority of research on albumin hydrogels has focused on bovine serum albumin (BSA), leaving human
Aida Maddahi et al.
BMC neuroscience, 9, 85-85 (2008-09-17)
MEK1/2 is a serine/threonine protein that phosphorylates extracellular signal-regulated kinase (ERK1/2). Cerebral ischemia results in enhanced expression of cerebrovascular contractile receptors in the middle cerebral artery (MCA) leading to the ischemic region. Here we explored the role of the MEK/ERK
Karine Loulier et al.
PLoS biology, 7(8), e1000176-e1000176 (2009-08-19)
During embryogenesis, the neural stem cells (NSC) of the developing cerebral cortex are located in the ventricular zone (VZ) lining the cerebral ventricles. They exhibit apical and basal processes that contact the ventricular surface and the pial basement membrane, respectively.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service