Skip to Content
Merck
All Photos(1)

Key Documents

P2287

Sigma-Aldrich

TWEEN® 20

viscous liquid, suitable for cell culture

Synonym(s):

Polysorbate 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

description

non-ionic

form

viscous liquid

mol wt

~1228

composition

lauric acid, ≥40% (balance primarily myristic, palmoyic, and stearic acids)

concentration

≥40.0%

technique(s)

cell culture | mammalian: suitable

refractive index

n20/D 1.468 (lit.)

CMC

0.06 mM (20-25°C)

transition temp

cloud point 76 °C

solubility

water: soluble

density

1.095 g/mL at 25 °C (lit.)
1.1 g/cm3 at 25 °C (lit.)

HLB

16.7

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tween 20 is a polyoxyethylene sorbitol ester that belongs to the polysorbate family. It is a nonionic detergent having a molecular weight of 1,225 daltons, assuming 20 ethylene oxide units, 1 sorbitol, and 1 lauric acid as the primary fatty acid. The ethylene oxide subunits are responsible for the hydrophilic nature of the surfactant, while the hydrocarbon chains provide the hydrophobic environment. Sorbitol forms the backbone ring to which the ethylene oxide polymers are attached.

Application

Non-ionic detergent
TWEEN 20 has been used in staining buffer for in situ hybridization. It has also been used to prevent non-specific adsorption of proteins to coverslip during sample preparation for FRET (Förster resonance energy transfer) experiments.
TWEEN 20 is a nonionic detergent widely used in biochemical applications. It has been used as an emulsifying agent for the preparation of stable oil-in-water emulsions. TWEEN 20 has been used in pre-extraction of membranes to remove peripheral proteins (used at 2% for extraction of membrane-bound proteins).
TWEEN 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%. TWEEN 20 can be used for lysing mammalian cells at a concentration of 0.005 to 0.5%.

Caution

TWEEN 20 may not be stable to autoclaving, particularly with metal cations in buffer solutions. Autoclaving is not recommended without testing for changes in properties. TWEEN 20 is heat sensitive and will darken when exposed to elevated temperatures. Polysorbates have been reported to be incompatible with alkalis, heavy metal salts, phenols, and tannic acid. They may also reduce the activity of many preservatives. Aqueous solutions of polysorbates undergo autoxidation during storage, with changes being catalyzed by light, increased temperature, and copper sulfate.

Preparation Note

TWEEN 20 is miscible in water (100mg/mL), yielding a clear, yellow solution. It is also miscible with alcohol, dioxane, and ethyl acetate. TWEEN 20 is insoluble in liquid paraffin and fixed oils.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

527.0 °F - Pensky-Martens closed cup

Flash Point(C)

275 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adverse morphological development in embryonic zebrafish exposed to environmental concentrations of contaminants individually and in mixture.
Kinch CD, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 175, 286-298 (2016)
Enoch Ming Wei Ng et al.
The Review of scientific instruments, 87(11), 115107-115107 (2016-12-03)
A method to measure the contact angle, which is indicative of wetting, using small liquid volumes dispensed directly on microplate wells is described and demonstrated. Experiments with enhanced green protein samples of volumes 4.4-6 μl showed no measured variance in
Conformations of a metastable SH3 domain characterized by smFRET and an excluded-volume polymer model.
Mazouchi A, et al.
Biophysical Journal, 110(7), 1510-1522 (2016)
Cassandra D Kinch et al.
Aquatic toxicology (Amsterdam, Netherlands), 175, 286-298 (2016-04-24)
Exposure to environmental contaminants has been linked to developmental and reproductive abnormalities leading to infertility, spontaneous abortion, reduced number of offspring, and metabolic disorders. In addition, there is evidence linking environmental contaminants and endocrine disruption to abnormal developmental rate, defects
Jeffrey Wimsatt et al.
BMC cancer, 16(1), 942-942 (2016-12-09)
Colorectal cancer is the second most common cause of cancer deaths for both men and women, and the third most common cause of cancer in the U.S. Toxicity of current chemotherapeutic agents for colorectal cancer, and emergence of drug resistance

Articles

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service