Skip to Content
Merck
All Photos(1)

Key Documents

MGVS100500

Medical Viscosity Standard

10.0 mPa.s (25.00°C ), pkg of 500 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Medical Viscosity Standards can be found on the Paragon Scientific webpage
These products contain antibacterial agent.
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 under UKAS accreditation

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service