Skip to Content
Merck
All Photos(1)

Documents

GG1

Sigma-Aldrich

Glycerol Gelatin

aqueous slide mounting medium

Synonym(s):

Glycerol jelly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

Quality Level

form

liquid

application(s)

hematology
histology

storage temp.

2-8°C

General description

Glycerol alters the hygroscopic property of gelatin and its permeability to water vapour. Glycerol Gelatin or glycerine jelly medium is an aqueous slide mounting media, used for mounting samples in microscopy and histology.

Application

Glycerol Gelatin has been used for mounting endometrial tissue and pancreatic sections for microscopic examination. It has also been used for the immobilization and mounting of helminth eggs for on chip image analysis.

Components

Prepared with gelatin, glycerol and phenol. Very similar in composition to Kaiser′s glycerol jelly.

Preparation Note

Product must be warmed to 55-60 °C before use. At ~37 °C the product returns to semi-solid state.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On-chip imaging of Schistosoma haematobium eggs in urine for diagnosis by computer vision
Linder E, et al.
PLoS Neglected Tropical Diseases, 7(12), e2547-e2547 (2013)
P F Lopez et al.
Investigative ophthalmology & visual science, 37(5), 855-868 (1996-04-01)
To determine the cellular origin and the vascular endothelial growth factor (VEGF) immunoreactivity of the nonvascular stromal cells in surgically excised age-related macular degeneration (ARMD)-associated choroidal neovascular membranes (CNVMs). Immunohistochemical analysis was performed on frozen sections of eight surgically excised
R G Erben
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 45(2), 307-313 (1997-02-01)
Methylmethacrylate (MMA) embedding of undecalcified bone biopsies is a technique widely used for bone histomorphometry. However, conventional MMA embedding causes almost complete loss of enzyme activity and protein antigenicity in the tissues. Recently, an MMA embedding technique has been reported
A Northern contaminant mixture impairs pancreas function in obese and lean JCR rats and inhibits insulin secretion in MIN6 cells
Mailloux R, et al.
Toxicology, 334(1), 81-93 (2015)
LABORATORY TECHNIQUES PRESERVATION AND PERMANENT MOUNTS
Bryophyte Ecology, 14(3), 035016-035016 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service