Skip to Content
Merck
All Photos(1)

Key Documents

O8015

Sigma-Aldrich

Organo/Limonene Mount

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

clear

density

0.840 g/cm3

application(s)

hematology
histology

greener alternative category

storage temp.

room temp

Application

Organo/Limonene Mount is a ready-to-use mounting medium is made with limonene, a natural product from orange peels. It is good for coverslipping stained tissues and cell smears that can be dehydrated with organic solvents. This mounting medium is suitable for organic solvent resistant immunohistochemistry (IHC) chromogens as well as an excellent choice for mounting H and E stained slides.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to toluene based mounting mediums.

Linkage

Organo/Limonene Mount is comparable to Clarion mounting medium.

Legal Information

Clarion is a trademark of Biomeda Corp.
Organo/Limonene Mount is a trademark of ImmunoBioScience Corp.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HDAC11 Suppresses the Thermogenic Program of Adipose Tissue via BRD2.
Bagchi R A, et al.
bioRxiv, 292011-292011 (2018)
Impaired Interoception in a Preclinical Model of Functional Dyspepsia.
Winston J H, et al.
Digestive Diseases and Sciences, 62(9), 2327-2337 (2017)
Daniel Chin Shiuan Lio et al.
Science advances, 6(22), eaaz6919-eaaz6919 (2020-06-12)
Transdermal drug delivery uses chemical, physical, or biochemical enhancers to cross the skin barrier. However, existing platforms require high doses of chemical enhancers or sophisticated equipment, use fragile biomolecules, or are limited to a certain type of drug. Here, we
Supplementation of specific carbohydrates results in enhanced deposition of chondrogenic-specific matrix during mesenchymal stem cell differentiation.
Trudy C W, et al.
Journal of Tissue Engineering and Regenerative Medicine, 15(5), 1261-1272 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service