Skip to Content
Merck
All Photos(1)

Documents

F0700000

Furosemide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfonyl)-4-chloro-2-([2-furanylmethyl]amino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

furosemide

manufacturer/tradename

EDQM

mp

220 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NS(=O)(=O)c1cc(C(O)=O)c(NCc2ccco2)cc1Cl

InChI

1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)

InChI key

ZZUFCTLCJUWOSV-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Furosemide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Inhibits ion co-transport in the kidney.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Case scenario: Hemodynamic management of postoperative acute kidney injury.
Matthieu Legrand et al.
Anesthesiology, 118(6), 1446-1454 (2013-04-06)
Maarten C van Loon et al.
Hearing research, 298, 17-26 (2013-01-31)
It is well known that spiral ganglion cells (SGCs) degenerate in hair-cell-depleted cochleas and that treatment with exogenous neurotrophins can prevent this degeneration. Several studies reported that, in addition, SGC size decreases after deafening and increases after neurotrophic treatment. The
Naoki Sato et al.
Circulation journal : official journal of the Japanese Circulation Society, 77(4), 944-951 (2013-03-19)
Hospitalized heart failure (HHF) is a critical issue in Japan. To improve its management and outcomes, the clinical features, in-hospital management, and outcomes should be analyzed to improve the guidelines for HHF. The acute decompensated heart failure syndromes (ATTEND) registry
Xin-Gui Peng et al.
Radiology, 269(3), 748-757 (2013-08-01)
To determine the relationship between renal lipid content and intrarenal oxygenation in diabetic nephropathy by using noninvasive chemical shift-selective (CSS) imaging and blood oxygen level-dependent (BOLD) magnetic resonance (MR) imaging. The study was approved by the institutional Committee on Animal
K M Ho et al.
Anaesthesia, 65(3), 283-293 (2010-01-21)
Furosemide, a potent loop diuretic, is frequently used in different stages of acute kidney injury, but its clinical roles remain uncertain. This review summarises the pharmacology of furosemide, its potential uses and side effects, and the evidence of its efficacy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service