Merck
Benefits and risks of furosemide in acute kidney injury.
Anaesthesia (2010-01-21)
K M Ho, B M Power
ABSTRACT
Furosemide, a potent loop diuretic, is frequently used in different stages of acute kidney injury, but its clinical roles remain uncertain. This review summarises the pharmacology of furosemide, its potential uses and side effects, and the evidence of its efficacy. Furosemide is actively secreted by the proximal tubules into the urine before reaching its site of action at the ascending limb of loop of Henle. It is the urinary concentrations of furosemide that determine its diuretic effect. The severity of acute kidney injury has a significant effect on the diuretic response to furosemide; a good 'urinary response' may be considered as a 'proxy' for having some residual renal function. The current evidence does not suggest that furosemide can reduce mortality in patients with acute kidney injury. In patients with acute lung injury without haemodynamic instability, furosemide may be useful in achieving fluid balance to facilitate mechanical ventilation according to the lung-protective ventilation strategy.
Product Description
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Furosemide
Furosemide for peak identification, European Pharmacopoeia (EP) Reference Standard