Skip to Content
Merck
All Photos(1)

Key Documents

F0040010

Fenbendazole impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Carbendazim, BCM, Methyl 2-benzimidazolecarbamate, Methyl benzimidazol-2-ylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O2
CAS Number:
Molecular Weight:
191.19
Beilstein:
649044
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fenbendazole

manufacturer/tradename

EDQM

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)Nc1nc2ccccc2[nH]1

InChI

1S/C9H9N3O2/c1-14-9(13)12-8-10-6-4-2-3-5-7(6)11-8/h2-5H,1H3,(H2,10,11,12,13)

InChI key

TWFZGCMQGLPBSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenbendazole impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 1B - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Christoph Thalhammer et al.
Nephrology (Carlton, Vic.), 20(2), 91-95 (2014-10-28)
Haemodynamic stability of patients during haemodialysis (HD) sessions is of pivotal importance and accurate determination of dry weight remains a challenge. Little information is available about central venous and aortic pressure during dialysis. In this pilot study we used a
Jae-Ran Lee et al.
Human mutation, 36(1), 69-78 (2014-09-30)
KIF1A is a neuron-specific motor protein that plays important roles in cargo transport along neurites. Recessive mutations in KIF1A were previously described in families with spastic paraparesis or sensory and autonomic neuropathy type-2. Here, we report 11 heterozygous de novo
Wojciech Dabrowski et al.
Clinical nephrology, 82(1), 41-50 (2014-06-03)
To analyze the effect and the time course of continuous veno-venous hemofiltration (CVVH) with net ultrafiltration (UF) on intra-abdominal pressure (IAP) body fluid volumes in septic shock patients with acute kidney injury (AKI). Patients were studied at baseline and after
Bin He et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(51), 18261-18266 (2014-12-10)
The androgen receptor (AR) is a key driver of prostate cancer (PC), even in the state of castration-resistant PC (CRPC) and frequently even after treatment with second-line hormonal therapies such as abiraterone and enzalutamide. The persistence of AR activity via
Przemyslaw Szafranski et al.
European journal of human genetics : EJHG, 23(2), 173-179 (2014-05-16)
Genomic copy-number variations (CNVs) constitute an important cause of epilepsies and other human neurological disorders. Recent advancement of technologies integrating genome-wide CNV mapping and sequencing is rapidly expanding the molecular field of pediatric neurodevelopmental disorders. In a previous study, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service