Skip to Content
Merck
All Photos(1)

Documents

E004000

Sigma-Aldrich

Kanamycin sulfate from Streptomyces kanamyceticus

Animal Component-free

Synonym(s):

Kanamycin A, Kanamycin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

Quality Level

form

powder

potency

≥750 μg per mg (dry basis)

impurities

≤5% Kanamycin B

SMILES string

OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1

InChI key

OOYGSFOGFJDDHP-KMCOLRRFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Kanamycin sulfate is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100 mg/mL.

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance:Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.
Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance:Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Caution

Solutions are stable at 37°C for approximately 5 days. Aqueous stock solutions can be stored at 2-8°C for long term storage.

Preparation Note

Kanamycin sulfate is soluble in water at 50 mg/mL, yielding a clear solution. It is practically insoluble in alcohol, acetone, chloroform, ether and ethyl acetate. A 1% solution in water has a pH of 6.5 to 8.5. Sterile solutions can be prepared by a sterile filtration, through a .2μm filter.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsutaka Kitano et al.
Antimicrobial agents and chemotherapy, 58(8), 4795-4803 (2014-06-11)
Highly pathogenic avian influenza A (H5N1) viruses cause severe and often fatal disease in humans. We evaluated the efficacy of repeated intravenous dosing of the neuraminidase inhibitor peramivir against highly pathogenic avian influenza virus A/Vietnam/UT3040/2004 (H5N1) infection in cynomolgus macaques.
Jiansheng Xiao et al.
International journal of molecular medicine, 43(2), 1094-1104 (2018-11-30)
Modulation of differentiation of dendritic cells (DCs), which are derived from bone marrow cells, may influence their maturation and consequently regulate their ability to present antigens to alloreactive T lymphocytes. B lymphocyte‑induced maturation protein‑1 (Blimp1) is a master regulator of
Fakhri Jeddi et al.
Antimicrobial agents and chemotherapy, 58(8), 4866-4874 (2014-06-11)
Antimonials remain the first-line treatment for the various manifestations of leishmaniasis in most areas where the disease is endemic, and increasing cases of therapeutic failure associated with parasite resistance have been reported. In this study, we assessed the molecular status
Bin Ye et al.
Autophagy, 15(6), 998-1016 (2019-02-02)
Macroautophagy/autophagy dysfunction is associated with many neurodegenerative diseases. TFEB (transcription factor EB), an important molecule that regulates lysosomal and autophagy function, is regarded as a potential target for treating some neurodegenerative diseases. However, the relationship between autophagy dysfunction and spiral
Atef Allam et al.
Journal of immunology (Baltimore, Md. : 1950), 193(2), 871-878 (2014-06-11)
The role of the TNF family member CD70 in adaptive T cell responses has been intensively studied, but its function in innate responses is still under investigation. In this study, we show that CD70 inhibits the early innate response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service